    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CDY1/2/L1/L2 Antibodies
          Antibodies Inc
          CDY1/2/L1/L2 Monoclonal Antibody (N143/36), FL594
          View all (5) CDY1/2/L1/L2 antibodies
          Tech Support
          Cite CDY1/2/L1/L2 Monoclonal Antibody (N143/36), FL594
          Additional Information:
          {{banner.description}}
          CDY1/2/L1/L2 Monoclonal Antibody (N143/36), FL594
          Product Details
          75-202-FL594
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N143/36
          Immunogen
          Fusion protein amino acids 6-66 (Chromo domain) of human CDY1 (accession number Q9Y6F8) produced recombinantly in E. Coli.
          View immunogen
          Conjugate
          FL594 FL594 FL594
          • Unconjugated
          • FL490
          • FL550
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          594/615 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          CDY1, CDY2, CDYL1, and CDYL2 are members of the chromodomain Y-like (CDY) family of proteins, which are involved in chromatin remodeling and gene regulation. CDY1 and CDY2 are Y-linked genes specifically important for spermatogenesis, while CDYL1 and CDYL2 are autosomal and have broader roles in regulating chromatin structure and gene expression in various tissues. These proteins contribute to processes such as development, differentiation, and cellular response to environmental signals, and their dysfunction can be linked to reproductive and developmental disorders.
          Bioinformatics
          Protein Aliases: CDY1; CDY1/CDY2A/CDY1L1/CDYL2; CDY2; chromodomain protein, Y chromosome, 1; chromodomain protein, Y chromosome, 2; chromodomain protein, Y-linked, 1; chromodomain protein, Y-linked, 2A; testis-specific chromodomain protein on Y; Testis-specific chromodomain protein Y 1; Testis-specific chromodomain protein Y 2; unnamed protein product; Y chromosome chromodomain protein 2A
          View more View less
          Gene Aliases: CDY; CDY1; CDY1A; CDY1B; CDY2; CDY2A; CDY2B
          View more View less
          UniProt ID: (Human) Q9Y6F8, (Human) Q9Y6F7
          View more View less
          Entrez Gene ID: (Human) 9085, (Human) 9426
          View more View less
          Function(s)
          transcription corepressor activity histone acetyltransferase activity transferase activity transferase activity, transferring acyl groups H3K27me3 modified histone binding peptide-lysine-N-acetyltransferase activity histone H3K9me2/3 reader activity
          Process(es)
          chromatin organization chromatin remodeling spermatogenesis negative regulation of transcription, DNA-templated
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline