          • Primary Antibodies ›
          • PEX5L Antibodies
          Antibodies Inc
          TRIP8b (Exon 4) Monoclonal Antibody (N212/3), FL550
          View all (29) PEX5L antibodies
          Tech Support
          Cite TRIP8b (Exon 4) Monoclonal Antibody (N212/3), FL550
          Additional Information:
          {{banner.description}}
          TRIP8b (Exon 4) Monoclonal Antibody (N212/3), FL550
          Product Details
          75-208-FL550
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N212/3
          Immunogen
          Fusion protein amino acids 1-192 (exon 1a, exon 4 and constant region) of rat TRIP8b (accession number Q925N3) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL550 FL550 FL550
          • Unconjugated
          • FL490
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          550/575 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Target Information
          Pex5l is an accessory subunit of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels, regulating their cell-surface expression and cyclic nucleotide dependence. Research studies have shown that Pex5l is expressed in the brain, testis and pancreas. Diseases associated with PEX5L include Rhizomelic Chondrodysplasia Punctata, Type 5 and Rhizomelic Chondrodysplasia Punctata, Type 2.
          Bioinformatics
          Protein Aliases: HCN channel auxiliary subunit; peroxin 2; Peroxin-5-related protein; peroxisomal targeting signal 1 receptor-like; Peroxisome biogenesis factor 5-like; PEX2-related protein; PEX5-like protein; PEX5-related protein; Pex5p-related protein; Pex5Rp; Tetratricopeptide repeat-containing Rab8b-interacting protein; TPR-containing Rab8b-interacting protein; unnamed protein product
          View more View less
          Gene Aliases: 1700016J08Rik; Pex2; PEX5L; PEX5R; PEX5RP; PXR2; PXR2B; TRIP8b; Trjip8b
          View more View less
          UniProt ID: (Human) Q8IYB4, (Rat) Q925N3, (Mouse) Q8C437
          View more View less
          Entrez Gene ID: (Human) 51555, (Rat) 286937, (Mouse) 58869
          View more View less
          Function(s)
          peroxisome targeting sequence binding peroxisome matrix targeting signal-1 binding protein binding small GTPase binding intracellular cyclic nucleotide activated cation channel activity Rab GTPase binding membrane trafficking regulatory protein
          Process(es)
          protein import into peroxisome matrix, docking cation transport protein import into peroxisome matrix regulation of membrane potential regulation of cAMP-mediated signaling regulated exocytosis maintenance of protein location positive regulation of corticotropin secretion cation transmembrane transport
          Contact tech support
          オンラインオーダー Plus Icon Minus Icon
          • オンラインオーダー可能な製品
          • オンラインオーダーに関する個人情報の取扱いについて
          • 特定商取引法に基づく表記
          • 返品について
          • 表示価格について
          • 配送料について
          サポート Plus Icon Minus Icon
          • FAQ、サポート情報
          • お問い合わせ一覧
          • サポートセンター
          • ドキュメント、証明書
          • 研究情報公開
          • Webサイトの問題を報告​
          リソース Plus Icon Minus Icon
          • ラーニングセンター
          • キャンペーン
          • イベント、セミナー
          • Blog
          • 企業情報
          • Social Responsibility (CSR)
          • お客さま体験の向上
          • 採用情報 採用情報
          • ニュース
          • 商標
          • ポリシーと通知
          • PPD
          • 標準販売条件
          • 個人情報保護方針
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline