    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • Kir6.2 (KCNJ11) Antibodies
          Antibodies Inc
          Kir6.2 Potassium Channel Monoclonal Antibody (N363/71), FL650
          View all (18) Kir6.2 (KCNJ11) antibodies
          Tech Support
          Cite Kir6.2 Potassium Channel Monoclonal Antibody (N363/71), FL650
          Additional Information:
          {{banner.description}}
          Kir6.2 Potassium Channel Monoclonal Antibody (N363/71), FL650
          Product Details
          75-393-FL650
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N363/71
          Immunogen
          Fusion protein amino acids 345-390 of rat Kir6.2 (accession number P70673) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL650 FL650 FL650
          • Unconjugated
          • FL490
          • FL550
          • FL594
          • Request custom conjugation
          Excitation/Emission Max
          655/676 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: TARQLDEDRS LLDALTLASS RGPLRKRSVA VAKAKPKFSI SPDSLS
          Target Information
          This receptor is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. Can be blocked by extracellular barium. Subunit of ATP-sensitive potassium channels (KATP). Can form cardiac and smooth muscle-type KATP channels with ABCC9. KCNJ11 forms the channel pore while ABCC9 is required for activation and regulation.
          Bioinformatics
          Protein Aliases: AI842722; ATP-sensitive inward rectifier potassium channel 11; AW491124; BIR; Inward rectifier K(+) channel Kir6.2; IRK11; KATP; KATP channel; MGC133230; Potassium channel, inwardly rectifying subfamily J member 11; potassium inwardly rectifying channel, subfamily J, member 11; potassium inwardly-rectifying channel subfamily J member 11
          View more View less
          Gene Aliases: Kcnj11; Kir6.2; mBIR
          View more View less
          UniProt ID: (Rat) P70673, (Mouse) Q61743
          View more View less
          Entrez Gene ID: (Rat) 83535, (Mouse) 16514
          View more View less
          Function(s)
          voltage-gated potassium channel activity protein binding ATP binding protein C-terminus binding ATP-activated inward rectifier potassium channel activity G-protein activated inward rectifier potassium channel activity ankyrin binding heat shock protein binding ion channel binding inward rectifier potassium channel activity voltage-gated ion channel activity potassium ion binding ion channel transporter
          Process(es)
          response to ischemia glucose metabolic process potassium ion transport potassium ion import response to estradiol response to ATP response to testosterone regulation of ion transmembrane transport regulation of membrane potential response to drug negative regulation of insulin secretion regulation of insulin secretion neurological system process cellular response to nicotine cellular response to glucose stimulus cellular response to tumor necrosis factor potassium ion transmembrane transport positive regulation of cation channel activity transport ion transport
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline