  • Applications
    • Real-Time PCR
    • Cloning
    • Protein Biology
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • Kir6.2 (KCNJ11) Antibodies
          Antibodies Inc
          Kir6.2 Potassium Channel Monoclonal Antibody (N363/71), FL490
          View all (18) Kir6.2 (KCNJ11) antibodies
          Tech Support
          Cite Kir6.2 Potassium Channel Monoclonal Antibody (N363/71), FL490
          Additional Information:
          {{banner.description}}
          Kir6.2 Potassium Channel Monoclonal Antibody (N363/71), FL490
          Product Details
          75-393-FL490
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          N363/71
          Immunogen
          Fusion protein amino acids 345-390 of rat Kir6.2 (accession number P70673) produced recombinantly in E. Coli.
          3D Epitope / Immunogen
          Conjugate
          FL490 FL490 FL490
          • Unconjugated
          • FL550
          • FL594
          • FL650
          • Request custom conjugation
          Excitation/Emission Max
          491/515 nm
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Product Specific Information
          For maximum recovery of product, centrifuge the vial prior to removing the cap.
          Sequence of this protein is as follows: TARQLDEDRS LLDALTLASS RGPLRKRSVA VAKAKPKFSI SPDSLS
          Target Information
          This receptor is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. Can be blocked by extracellular barium. Subunit of ATP-sensitive potassium channels (KATP). Can form cardiac and smooth muscle-type KATP channels with ABCC9. KCNJ11 forms the channel pore while ABCC9 is required for activation and regulation.
          Bioinformatics
          Protein Aliases: AI842722; ATP-sensitive inward rectifier potassium channel 11; AW491124; BIR; Inward rectifier K(+) channel Kir6.2; IRK11; KATP; KATP channel; MGC133230; Potassium channel, inwardly rectifying subfamily J member 11; potassium inwardly rectifying channel, subfamily J, member 11; potassium inwardly-rectifying channel subfamily J member 11
          View more View less
          Gene Aliases: Kcnj11; Kir6.2; mBIR
          View more View less
          UniProt ID: (Rat) P70673, (Mouse) Q61743
          View more View less
          Entrez Gene ID: (Rat) 83535, (Mouse) 16514
          View more View less
          Function(s)
          voltage-gated potassium channel activity protein binding ATP binding protein C-terminus binding ATP-activated inward rectifier potassium channel activity G-protein activated inward rectifier potassium channel activity ankyrin binding heat shock protein binding ion channel binding inward rectifier potassium channel activity voltage-gated ion channel activity potassium ion binding ion channel transporter
          Process(es)
          response to ischemia glucose metabolic process potassium ion transport potassium ion import response to estradiol response to ATP response to testosterone regulation of ion transmembrane transport regulation of membrane potential response to drug negative regulation of insulin secretion regulation of insulin secretion neurological system process cellular response to nicotine cellular response to glucose stimulus cellular response to tumor necrosis factor potassium ion transmembrane transport positive regulation of cation channel activity transport ion transport
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          • Documents and Certificates
          • Study information disclosure
          • Promotions
          • Events and Seminars
          • Blog
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          Japan flag icon
          Japan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline