    • Alle Produktkategorien anzeigen
  • Anwendungen
    • Bioprocessing
    • Zellkultur und Transfektion
    • Zell- und Gentherapie
    • Chromatographie
    • Molekulare Testung
    • Digitale Lösungen
    • Extraktion und Analyse von DNA und RNA
    • Spektroskopie, Element- und Isotopenanalyse
    • Alle Anwendungen und Verfahren anzeigen
  • Service
    • 360° CDMO- und CRO-Services
    • CDMO-Services
    • CRO-Services
    • Kundenspezifische Services
    • Finanzierung und Leasing
    • Geräteservice
    • Laborinformatik
    • OEM und kommerzielle Bereitstellung
    • Schulungsdienstleistungen
    • Unity Lab Services
    • Alle Services anzeigen
  • Hilfe und Support
    • Für ein Konto registrieren
    • Bestellinformationen
    • Gerätesupport
    • Center für technischen Support
    • Lerncenter
    • Alle Themen für Hilfe und Support anzeigen
  • Häufigste Themen
      Cell Culture Plastics
  • Für wen wir arbeiten
    • Biotechnologie
    • Biopharma
    • CDMO
    • Labordiagnostik
    • Angewandte Wissenschaften und Industrie
  • Sonderaktionen
  • Kontaktieren Sie uns
  • Eilbestellung
  • Status und Nachverfolgung von Bestellungen
  • Dokumente und Zertifikate
Thermo Fisher Scientific Logo
Search
Suchen in Alle
Search button
          • Auftragsstatus
          • Eilbestellung
          • Aktionen
          • Anmeldung
            Anmeldung
            Sie haben noch kein Konto? Registrieren Sie sich hier
            • Konto
            • Bestellungen
            • Connect: Labor, Daten, Apps
            • Spezialanfertigungen und Projekte
            • Services Central
          • Primary Antibodies ›
          • ZRANB3 Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Bethyl Laboratories
          ZRANB3 Polyclonal Antibody
          View all (5) ZRANB3 antibodies
          Tech Support
          Cite ZRANB3 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          Product Details
          A303-033A-M
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Product Specifications
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Between 1029 and 1079
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.10 mg/mL
          Amount
          10 µg
          Storage conditions
          4°C
          Shipping conditions
          Wet ice
          Product Specific Information
          The recommended shelf life for this product is 1 year from date of receipt.
          Target Information
          DNA annealing helicase and endonuclease required to maintain genome stability at stalled or collapsed replication forks by facilitating fork restart and limiting inappropriate recombination that could occur during template switching events (PubMed:21078962, PubMed:22704558, PubMed:22705370, PubMed:22759634, PubMed:26884333). Recruited to the sites of stalled DNA replication by polyubiquitinated PCNA and acts as a structure-specific endonuclease that cleaves the replication fork D-loop intermediate, generating an accessible 3'-OH group in the template of the leading strand, which is amenable to extension by DNA polymerase (PubMed:22759634). In addition to endonuclease activity, also catalyzes the fork regression via annealing helicase activity in order to prevent disintegration of the replication fork and the formation of double-strand breaks (PubMed:22705370, PubMed:22704558). [UniProt]
          Contact tech support
          Bestellung Plus Icon Minus Icon
          • Auftragsstatus
          • Hilfe zu Bestellungen
          • Eilbestellung
          • Versorgungszentrum
          • Elektronische Beschaffung
          • Preis- und Frachtregeln
          Support Plus Icon Minus Icon
          • Hilfe und Support
          • Kontaktieren Sie uns
          • Center für technischen Support
          • Dokumente und Zertifikate
          • Meldung von Problemen auf der Webseite
          Ressourcen Plus Icon Minus Icon
          • Lerncenter
          • Sonderaktionen
          • Veranstaltungen und Webinare
          • Soziale Netzwerke
          Über Thermo Fisher Plus Icon Minus Icon
          • Über uns Über uns
          • Karriere Karriere
          • Investoren Investoren
          • News News
          • Soziale Verantwortung Soziale Verantwortung
          • Marken
          • Lieferkettensorgfaltspflichtengesetz
          • Richtlinien und Hinweise
          Unser Portfolio Plus Icon Minus Icon
          • Privacy Information Center
          • Impressum
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline