    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
          • Primary Antibodies ›
          • MICA/MICB Antibodies
          Proteintech
          MICA/MICB Monoclonal Antibody (6D4), APC
          View all (14) MICA/MICB antibodies
          Tech Support
          Cite MICA/MICB Monoclonal Antibody (6D4), APC
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          MICA/MICB Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          MICA/MICB Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          MICA/MICB Antibody (APC-65161) in Flow
          1x10^6 HeLa cells were surface stained with 5 µL APC Anti-Human MICA/MICB (APC-65161, Clone: 6D4) or APC Mouse IgG2a Isotype Control. Cells were not fixed. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MICA/MICB Antibody in Flow Cytometry (Flow)
          MICA/MICB Antibody in Flow Cytometry (Flow)
          MICA/MICB Monoclonal Antibody (6D4), APC
          Product Details
          APC-65161
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          6D4
          Immunogen
          Transfected C1R cells expressing MICA
          3D Epitope / Immunogen
          Conjugate
          APC APC APC
          • Biotin 
          • Cardinal Red
          • CoraLite Plus 488
          • CoraLite Plus 647
          • CoraLite Plus 750
          • CoraLitePlus 555
          • FITC Plus
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          651/660 nm View spectra spectra
          Form
          Liquid
          Concentration
          5 µL/Test
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS with 0.5% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          MICA encodes the higly polymorphic MHC (HLA) class I chain-related gene A. The protein product is expressed on the cell surface, although unlike canonical class I molecules does not seem to associate with beta-2-microglobulin. It is thought that MICA functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: cell stress-regulated membrane protein; HLA class I antigen; ligand to the NKG2D receptor; major histocompatibility complex class I chain-related protein A; MHC class I antigen-related protein B; MHC class I chain-related protein A; MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence A; MHC class I polypeptide-related sequence B; MHC class I related chain A; MHC class I related sequence A; MHC class I-like molecule PERB11.2-IMX; MIC-A; MIC-B; sMICA; sMICB; soluble MICA; soluble MICB; stress inducible class I homolog; unnamed protein product
          View more View less
          Gene Aliases: MIC-A; MICA; MICB; PERB11.1; PERB11.2
          View more View less
          UniProt ID: (Human) Q95IY7, (Human) Q29980
          View more View less
          Entrez Gene ID: (Human) 100507436, (Human) 4277
          View more View less
          Function(s)
          protein binding beta-2-microglobulin binding natural killer cell lectin-like receptor binding receptor agonist activity
          Process(es)
          T cell mediated cytotoxicity adaptive immune response immune system process immune response to tumor cell immune response cellular response to DNA damage stimulus signal transduction response to heat negative regulation of defense response killing of cells of other organism negative regulation of natural killer cell activation natural killer cell mediated cytotoxicity defense response to bacterium negative regulation of natural killer cell mediated cytotoxicity gamma-delta T cell activation regulation of immune response defense response to virus immune response-activating cell surface receptor signaling pathway response to oxidative stress response to retinoic acid negative regulation of defense response to virus by host
          • Technical Support Centers
          • Find Documents and Certificates
          Brasil flag icon
          Brasil
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline