  • Applications
    • Bioprocesamiento
    • Cultivo celular y transfección
    • Terapia celular y génica
    • Cromatografía
    • Pruebas moleculares
    • Soluciones digitales
    • Extracción y análisis de ADN y ARN
    • Espectroscopía, análisis elemental y de isótopos
    • Ver todas las aplicaciones y técnicas
  • Servicios
    • Servicios 360° de CDMO y CRO
    • Servicios de CDMO
    • Servicios de CRO
    • Servicios personalizados
    • Servicios de leasing y financiación
    • Servicios de instrumentos
    • Informática de laboratorio
    • OEM y oferta comercial
    • Servicios de formación
    • Unity Lab Services
    • Ver todos los servicios
  • Ayuda y soporte técnico
    • Regístrese para obtener una cuenta
    • Cómo hacer un pedido
    • Asistencia para el instrumental
    • Centros de soporte técnico
    • Centros de formación
    • Vea todos los temas de ayuda y soporte técnico
      Cell Culture Plastics
  • A quiénes brindamos nuestros servicios
    • Industria biotecnológica
    • Sector biofarmacéutico
    • CDMO
    • Diagnósticos de laboratorio
    • Ciencias aplicadas e industriales
  • Promociones
  • Contacto
  • Pedido rápido
  • Estado del pedido y seguimiento
  • Documentos y certificados
Thermo Fisher Scientific Logo
Search
Buscar
Search button
          • Estado del pedido
          • Pedido rápido
            • Cuenta
            • Pedidos
            • Connect: laboratorio, datos, aplicaciones
            • Productos y proyectos personalizados
            • Services Central
          • Primary Antibodies ›
          • 6x His Tag Antibodies
          Proteintech
          6x His, His-Tag Monoclonal Antibody (1B7G5), Biotin
          View all (136) 6x His Tag antibodies
          Tech Support
          Cite 6x His, His-Tag Monoclonal Antibody (1B7G5), Biotin
          Additional Information:
          {{banner.description}}
          Product Details
          BIOTIN-66005
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          1:10-1:100
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          1B7G5
          Immunogen
          Peptide
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • APC 
          • CoraLite 594
          • CoraLite Plus 488
          • CoraLite Plus 647
          • CoraLite Plus 750
          • HRP
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          PBS, pH 7.3, with 50% glycerol, 0.5% BSA
          Contains
          0.05% ProClin 300
          Storage conditions
          -20°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          The 6x His tag is a synthetic oligo peptide consisting of 6 consecutive histidine residues (HHHHHH). The His tag is commonly expressed as a tag at either N-or C-terminal regions of recombinant proteins to allow isolation or purification by immobilized metal affinity chromatography. Epitope tagging is a technique in which a known epitope is fused to a recombinant protein using genetic engineering. By choosing a particular epitope and recombinant protein combination, epitope tagging makes it possible to detect proteins for which no antibody is available. His epitope tagged proteins that contain a stretch of histidine residues at the carboxyl or amino terminus are used for purification of recombinant proteins by means of metal chelate chromatography due to metal binding properties of histidine clusters on His tagged proteins. For example, the affinity of the histidine-tag motif to Ni2+ by chelation is strong and selective enough to enable purification of the protein to homogeneity by affinity chromatography on a Ni2+-NTA adsorbent. His tag-specific antibodies are used to facilitate detection or coimmunopreciptation of tagged proteins.
          Bioinformatics
          Protein Aliases: 6His tag; 6X His; His Tag; Histidine Tag
          Pedidos Plus Icon Minus Icon
          • Estado del pedido
          • Ayuda con el pedido
          • Pedido rápido
          • Centro de suministros
          • eProcurement (B2B)
          Asistencia Plus Icon Minus Icon
          • Ayuda y soporte técnico
          • Contacto
          • Centros de soporte técnico
          • Documentos y certificados
          • Informar de un problema del sitio web
          Recursos Plus Icon Minus Icon
          • Centros de formación
          • Promociones
          • Eventos y seminarios web
          • Redes sociales
          Acerca de Thermo Fisher Plus Icon Minus Icon
          • Acerca de nosotros Acerca de nosotros
          • Carreras profesionales Carreras profesionales
          • Inversores Inversores
          • Noticias Noticias
          • Responsabilidad social Responsabilidad social
          • Marcas registradas
          • Políticas y avisos
          Nuestra cartera Plus Icon Minus Icon
          Spain flag icon
          Spain
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline