Search
Bioss
FIGURE: 1 / 1
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The function of this gene has not yet been determined.
Protein Aliases: annexin 14; Annexin A10; Annexin-10; Annexin-14
Gene Aliases: ANX14; ANXA10; RGD1560956
UniProt ID: (Human) Q9UJ72, (Mouse) Q9QZ10
Entrez Gene ID: (Human) 11199, (Mouse) 26359, (Rat) 498622