  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • gamma-H2AX Antibodies
          Identifyn
          gamma-H2AX SRM Recombinant Rabbit Monoclonal Antibody (BLR014M), Alexa Fluor™ 680, Identifyn™
          View all (17) gamma-H2AX antibodies
          Tech Support
          Cite gamma-H2AX SRM Recombinant Rabbit Monoclonal Antibody (BLR014M), Alexa Fluor™ 680, Identifyn™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (8)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 8
          gamma-H2AX SRM Antibody (DC-000022) in ICC/IF
          Identifyn Detailed protocols can be found below under Documents & Downloads. Super Resolution - SIM2 Microscopy: HeLa cells were grown on acid-etched 1.5H cover glass to ~50% confluency. DNA damage was induced via treatment with 2 µM Etoposide-spiked complete growth media for 24 hours, followed by -20°C methanol fixation and blocked with Thermo Fisher Fish Serum Blocker (Product # 37527X3). The primary anti... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Western Blot (WB)
          gamma-H2AX SRM Recombinant Rabbit Monoclonal Antibody (BLR014M), Alexa Fluor™ 680, Identifyn™
          Product Details
          DC-000022
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:250-1:1,000
          -
          Immunocytochemistry (ICC/IF)
          1:250-1,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          ExpiCHO
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          BLR014M
          Immunogen
          Synthetic peptide corresponding to Human gamma H2AX (phospho Serine 139), referencing both Uniprot P16104 and Gene ID 3014.
          3D Epitope / Immunogen
          Conjugate
          Alexa Fluor™ 680 Alexa Fluor™ 680 Alexa Fluor™ 680
          • Unconjugated
          • Alexa Fluor 405 
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 555 
          • Alexa Fluor 594 
          • Alexa Fluor 647 
          • Alexa Fluor 750 
          • Request custom conjugation
          Excitation/Emission Max
          681/704 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          50 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 10% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Identifyn™ Primary and Secondary antibodies are extensively validated in Immunofluorescence assays by traditional as well as high resolution acquisition like Widefield SIM, Confocal, Airyscan and STORM Microscopy.
          Recommended dilution for traditional microscopy:
          Widefield - 1:250-1:1,000
          Widefield SIM - 1:250-1:1,000
          Confocal - 1:250-1:1,000
          Recommended dilution for Super Resolution microscopy:
          SIM (Structured Illumination Microscopy) - 1:1,000
          Single Molecule STORM (Stochastic Optical Reconstruction Microscopy) - 1:100-1:1,000
          Detailed protocols can be found below under Documents & Downloads.
          Target Information
          H2AX is a variant of the H2A protein family and is a component of the histone octomer in nucleosomes. H2AX is phosphorylated by kinases such as ataxia telangiectasia mutated (ATM) and ATM-Rad3-related (ATR) in the PI3K pathway. Gamma-H2AX is the first step in recruiting and localizing DNA repair proteins. DSBs can be induced by mechanisms such as ionizing radiation or cytotoxic agents and subsequently, gamma-H2AX foci quickly form. These foci represent the DSBs in a 1:1 manner and can be used as a biomarker for damage. The visualization of gamma-H2AX by can allow the assessment of DNA damage, related DNA damage proteins and DNA repair. Gamma-H2AX has applications in the detection of genomic damage caused by cytotoxic chemical agents and environmental and physical damage.
          Bioinformatics
          Protein Aliases: gamma H2A X; gamma H2A.X; gamma-H2A.X; gh2ax; H2A histone family member X; H2a/x; H2AFX; H2AX histone; Histone H2A.X; Histone H2AX; unnamed protein product
          View more View less
          Gene Aliases: H2A.X; H2A/X; H2AFX; H2AX
          View more View less
          UniProt ID: (Human) P16104
          View more View less
          Entrez Gene ID: (Human) 3014
          View more View less
          Function(s)
          DNA binding damaged DNA binding protein binding enzyme binding structural constituent of chromatin histone binding protein heterodimerization activity chromatin-protein adaptor activity
          Process(es)
          DNA damage checkpoint double-strand break repair via homologous recombination DNA repair double-strand break repair DNA recombination nucleosome assembly cellular response to DNA damage stimulus spermatogenesis response to ionizing radiation heterochromatin assembly positive regulation of DNA repair meiotic cell cycle protein K63-linked ubiquitination positive regulation of double-strand break repair via homologous recombination protein localization to site of double-strand break
          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline