    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • gamma-H2AX Antibodies
          Identifyn
          gamma-H2AX SRM Recombinant Rabbit Monoclonal Antibody (BLR014M), Alexa Fluor™ 405, Identifyn™
          View all (17) gamma-H2AX antibodies
          Questions & Answers
          Cite gamma-H2AX SRM Recombinant Rabbit Monoclonal Antibody (BLR014M), Alexa Fluor™ 405, Identifyn™
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (10)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 10
          gamma-H2AX SRM Antibody (DC-000040) in ICC/IF
          Identifyn Detailed protocols can be found below under Documents & Downloads. Confocal Microscopy: HeLa cells were grown on acid-etched 1.5H cover glass to ~50% confluency. DNA damage was induced via treatment with 2 µM Etoposide-spiked complete growth media for 24 hours, followed by -20°C methanol fixation and blocked with Thermo Fisher Fish Serum Blocker (Product # 37527X3). The primary antibody, Rabbit an... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Immunocytochemistry (ICC/IF)
          gamma-H2AX SRM Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          gamma-H2AX SRM Recombinant Rabbit Monoclonal Antibody (BLR014M), Alexa Fluor™ 405, Identifyn™
          Product Details
          DC-000040
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:250-1:1,000
          -
          Immunocytochemistry (ICC/IF)
          1:250-1,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          ExpiCHO
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          BLR014M
          Immunogen
          Synthetic peptide corresponding to Human gamma H2AX (phospho Serine 139), referencing both Uniprot P16104 and Gene ID 3014.
          3D Epitope / Immunogen
          Conjugate
          Alexa Fluor™ 405 Alexa Fluor™ 405 Alexa Fluor™ 405
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 555 
          • Alexa Fluor 594 
          • Alexa Fluor 647 
          • Alexa Fluor 680 
          • Alexa Fluor 750 
          • Request custom conjugation
          Excitation/Emission Max
          401/422 nm View spectra spectra
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          50 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 10% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Identifyn™ Primary and Secondary antibodies are extensively validated in Immunofluorescence assays by traditional as well as high resolution acquisition like Widefield SIM, Confocal and Airyscan.
          Recommended dilution for traditional microscopy:
          Widefield - 1:250-1:1,000
          Widefield SIM - 1:250-1:1,000
          Confocal - 1:250-1:1,000
          Recommended dilution for Super Resolution microscopy:
          SIM (Structured Illumination Microscopy) - 1:1,000
          Detailed protocols can be found below under Documents & Downloads.
          Target Information
          H2AX is a variant of the H2A protein family and is a component of the histone octomer in nucleosomes. H2AX is phosphorylated by kinases such as ataxia telangiectasia mutated (ATM) and ATM-Rad3-related (ATR) in the PI3K pathway. Gamma-H2AX is the first step in recruiting and localizing DNA repair proteins. DSBs can be induced by mechanisms such as ionizing radiation or cytotoxic agents and subsequently, gamma-H2AX foci quickly form. These foci represent the DSBs in a 1:1 manner and can be used as a biomarker for damage. The visualization of gamma-H2AX by can allow the assessment of DNA damage, related DNA damage proteins and DNA repair. Gamma-H2AX has applications in the detection of genomic damage caused by cytotoxic chemical agents and environmental and physical damage.
          Bioinformatics
          Protein Aliases: gamma H2A X; gamma H2A.X; gamma-H2A.X; gh2ax; H2A histone family member X; H2AFX; H2AX histone; histone H2A.x; unnamed protein product
          View more View less
          Gene Aliases: H2A.X; H2A/X; H2AFX
          View more View less
          UniProt ID: (Human) P16104
          View more View less
          Entrez Gene ID: (Human) 3014
          View more View less
          Function(s)
          DNA binding damaged DNA binding protein binding enzyme binding structural constituent of chromatin histone binding protein heterodimerization activity chromatin-protein adaptor activity
          Process(es)
          DNA damage checkpoint double-strand break repair nucleosome assembly cellular response to DNA damage stimulus response to ionizing radiation heterochromatin assembly positive regulation of DNA repair protein localization to site of double-strand break
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch