    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • c-Kit (CD117) Antibodies
          Invitrogen
          CD117 (c-Kit) Monoclonal Antibody (kit2c75), PE
          View all (178) c-Kit (CD117) antibodies
          Tech Support
          Cite CD117 (c-Kit) Monoclonal Antibody (kit2c75), PE
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          Product Details
          MA5-28805
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.2 µg/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Chicken
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          kit2c75
          Immunogen
          Sp/chkit4A4 cells (Sp2/0 cells transfected with chicken c-Kit cDNA)
          Conjugate
          PE PE PE
          • Unconjugated
          • Biotin 
          • FITC
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.5, with proprietary stabilizer
          Contains
          <0.1% sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2745764
          Target Information
          KIT (c-KIT) is a proto-oncogene and a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). KIT was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. KIT together with its ligand regulates growth and activation of a variety of hemopoietic and non-hemopoietic cells. Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. Recently, deregulation of the KIT receptor TK by the prevalent activation loop mutation D816V has served as a focal point in therapeutic strategies aimed at curbing neoplastic mast cell growth. c-Kit is expressed in hematopoietic stem cells, germ cells, mast cells and gastrointestinal tract cajal cells. Upon binding of its ligand stem cell factor (SCF), c-kit dimerizes, resulting in receptor activation and autophosphorylation of various tyrosine residues including tyrosine 703 located on the cytoplasmic domain of the receptor. This modification allows docking of Grb2 and activation of the Ras/ERK signaling pathway. SCF/c-kit can activate multiple downstream signaling pathways including PI3K, PLC-gamma and JAK/STAT. c-kit receptor activation is essential for hematopoiesis, stem cell maintenance and gametogenesis.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: c-kit; ckit; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog
          View more View less
          Gene Aliases: SCFR
          View more View less
          UniProt ID: (Chicken) Q08156
          View more View less
          Entrez Gene ID: (Chicken) 378783
          View more View less
          Function(s)
          protease binding transmembrane receptor protein tyrosine kinase activity stem cell factor receptor activity ATP binding cytokine binding SH2 domain binding protein homodimerization activity metal ion binding transmembrane signal receptor
          Process(es)
          activation of MAPK activity ovarian follicle development hematopoietic progenitor cell differentiation myeloid progenitor cell differentiation lymphoid progenitor cell differentiation immature B cell differentiation dendritic cell cytokine production mast cell chemotaxis glycosphingolipid metabolic process inflammatory response transmembrane receptor protein tyrosine kinase signaling pathway multicellular organism development spermatid development positive regulation of cell proliferation regulation of cell shape male gonad development response to radiation positive regulation of gene expression peptidyl-tyrosine phosphorylation lamellipodium assembly B cell differentiation T cell differentiation erythrocyte differentiation melanocyte differentiation positive regulation of cell migration actin cytoskeleton reorganization mast cell cytokine production positive regulation of kinase activity embryonic hemopoiesis ectopic germ cell programmed cell death intracellular signal transduction hematopoietic stem cell migration megakaryocyte development Fc receptor signaling pathway Kit signaling pathway erythropoietin-mediated signaling pathway positive regulation of tyrosine phosphorylation of STAT protein negative regulation of programmed cell death mast cell degranulation positive regulation of MAP kinase activity positive regulation of JAK-STAT cascade protein autophosphorylation digestive tract development stem cell differentiation detection of mechanical stimulus involved in sensory perception of sound positive regulation of sequence-specific DNA binding transcription factor activity mast cell differentiation cellular response to thyroid hormone stimulus melanocyte migration melanocyte adhesion positive regulation of vascular smooth muscle cell differentiation
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-5bc99cc877-v966v:80/100.66.72.244:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0