Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • Unity Lab Services
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          • Isotype Controls ›
          • Phalloidin Isotype Controls
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          Phalloidin, DyLight™ 633

          View all Phalloidin Controls

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support
          Datasheet
          Certificates
          Tech Support

          Cite Phalloidin, DyLight™ 633

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Testing Data (5)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phalloidin Control (21840) in ICC/IF

          Immunofluorescent analysis of Phalloidin (magenta) and alpha-Tubulin (green) in NIH 3T3 cells. Formalin fixed cells were permeabilized with 0.1% Triton X-100 in PBS for 10 minutes at room temperature and blocked with 2% BSA (Product # 37525) in PBS + 0.1% Triton X-100 for 30 minutes at room temperature. Cells were probed with an alpha-Tubulin monoclonal antibody (Product # MA1-19162) at a dilution of 1:1000... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)
          Phalloidin Control in Immunocytochemistry (ICC/IF)

          Product Details

          21840

          Applications
          Tested Dilution

          Immunohistochemistry (IHC)

          Assay-dependent

          Immunocytochemistry (ICC/IF)

          1-5 units/mL
          Product Specifications

          Type

          Control

          Conjugate

          DyLight™ 633 DyLight™ 633 DyLight™ 633

          Excitation/Emission Max

          620/649 nm View spectra spectra

          Form

          Lyophilized

          Concentration

          300 U/mL

          Storage conditions

          -20°C

          Shipping conditions

          Ambient

          RRID

          AB_2532161

          Product Specific Information

          Format: 300 units of lyophilized DyLight 633-Phalloidin (300 units/mL) in methanol. Prepare stock solution by adding 1000 µL of pure methanol to the vial and gently mix.

          21840 has been successfully used in immunofluorescence and immunohistochemistry. DyLight 633-Phalloidin has an excitation/emission of 636/658 nm and molecular weight of 1506.77 g/mole.

          DyLight 633-Phalloidin Stock Solution is prepared in methanol, and is also soluble in DMF, or DMSO. The unused Stock Solution should be promptly stored at -20°C in a foil pouch with desiccant, and protected from light. The stock solutions are stable for at least one year. Staining cells with some of the DyLight Phalloidin conjugates may require the use of a higher concentration of the phalloidin conjugate; therefore to minimize the amount of methanol added to the cells, the vial contents could be dissolved in 0.5 mL methanol to yield a final concentration of 600units/mL.

          Working solution of 1unit/mL (per 96-well plate) can be prepared by diluting 20 µL of the Stock Solution in 5.98 mL of PBS and mixing well. Typical staining procedure adds 50 µL of Working Solution (i.e. 1 to 5 unit/mL) to a each well. Incubate cells in the dark for 30 minutes at room temperature (optimal staining times varies from 10 minutes to 3 hours depending on cell type). Aspirate and wash cells three times in PBS after incubation. If desired, probe with specific primary antibodies followed by secondary antibodies conjugated to any compatible fluorophores before using DyLight 633-Phalloidin conjugate.

          Target Information

          Phalloidin is a bicyclic peptide that belongs to a family of toxins isolated from the deadly Amanita phalloides 'death cap' mushroom and is commonly used as a counterstain (similar to DAPI or Hoechst) in cell biology and histology imaging applications to selectively label F-actin in fixed cells, permeabilized cells, and cell-free experiments. Labeled phalloidin conjugates have similar affinity for both large and small filaments and bind in a stoichiometric ratio of about one phallotoxin per actin subunit in both muscle and non-muscle cells. Phalloidins reportedly do not bind to monomeric G-actin, unlike some antibodies against actin. The dynamics of the actin polymerization in cells are important for a variety of cellular processes from cell motility to cell shape, from muscular contraction to cytokinesis, and more.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          View product citations for antibody 21840 on CiteAb

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline