Search
The Thermo Scientific Phenom XL G3 Desktop SEM offers a range of features to help you work more efficiently. From streamlined day-to-day operation to more predictable maintenance, it’s designed to help you focus on your work.
Cerium hexaboride (CeB6) crystals serve as thermionic electron sources, providing exceptional image quality with improved resolution and contrast. These durable electron sources significantly outlast traditional tungsten filaments, with a lifespan exceeding 30 times longer. Unlike tungsten filaments, CeB6 sources degrade gradually rather than experiencing sudden failures, so you can plan replacements during regular maintenance.
This system expands your research horizons thanks to a larger stage that can accommodate samples up to 100 mm x 100 mm. In fact, it can accept large samples and multiple samples all in one fast load, helping you maintain sample integrity by eliminating the need to move your samples between different instruments.
Automation is built into all Thermo Scientific SEMs, and the Phenom XL G3 Desktop SEM is no different. It automates various stages of your routine workflow, delivering accurate, reproduceable results with an average time to image of 60 seconds. It’s a proven way to streamline your operations and recoup valuable time.
The Phenom XL G3 Desktop SEM is optimized for both routine analysis and quality control sampling. Its non-reactive environment helps you study a wide range of reactive materials, including highly volatile samples like solid-state lithium batteries and steel.
We know the importance of ensuring consistent quality at an affordable price in industrial research and analysis. That’s why the Phenom XL G3 Desktop SEM was designed to provide the insights and information you need to discover failures earlier and adjust your production processes faster.
| Phenom XL G3 Desktop SEM | |
Sample size |
|
| Source type | Cerium hexaboride (CeB6) |
| Max. resolution | <10 nm_ |
| Detectors and signals |
|
Software options and accessories |
|
| Footprint | 93 (w) x 293 (d) x 290 (h) mm, 82 kg |
| Acceleration voltage range | 2 kV 5 kV to 20 kV. The low vac range is 0.1 Pa – 10 Pa – 60 Pa |
| Low vacuum mode (stating pressure ranges) | 0.1 Pa – 1 Pa – 60 Pa |