Reproducible results with powerful microarray analysis
Applied Biosystems microarray instruments provide a fully integrated platform for processing CytoScan arrays for cytogenetics research. Our instruments combine advanced technology with user-friendly features for microarray scanning needs to dramatically improve throughput and efficiency in your genetic analysis. 
Workflow
Thermo Fisher Scientific offers powerful microarray solutions for postnatal clinical research that are powered with robust workflows that make it easy to obtain consistent, reliable, and high-quality results. 
Our complete microarray research platform, the CytoScan Cytogenetics Suite, includes hybrid-SNP arrays, automated and manual target preparation options, fully kitted reagents, the GeneChip System 3000 instrument for array processing, and Chromosome Analysis Suite (ChAS) software for data interpretation and reporting. These tools support laboratories to help maximize operational time. 
Product overview
GeneChip System 3000 instrument platform
A fully integrated platform with advanced design and automation to enable high resolution scanning with minimized hands-on time.
This comprehensive system includes GeneChip Scanner 3000 7G, GeneChip Fluidics Station 450, GeneChip Hybridization Oven 645, and a powerful computer workstation with instrument control software. 
Key highlights of GeneChip System 3000 are:
  • Compact size for better space utilization
  • High-resolution scanning of 0.51–2.5 μm pixelations, automatically selected by array type
  • Optimal image uniformity and collection efficiency across entire scan area with proprietary Flying Objective lens technology
  • No laser drift and reduced scanner-to-scanner variability
  • Automatic adjustment of residual arc correction and x-linearity
  • Compatible with the GeneChip AutoLoader for complete walk-away scanning of up to 48 arrays at a time
GeneTitan MC Fast Scan Instrument
The GeneTitan MC Fast Scan Instrument is a hands-free, automated solution for analyzing genome-wide SNP genotyping using the CytoScan HT-CMA array plate. It automates array processing from target hybridization to data generation by combining a hybridization oven, fluidics processing, and state-of-the art imaging device into a single bench-top instrument.
Key features are: 
  • Scalable—meets both medium and high-throughput needs; enables fast time to data with fewer resources 
  • Efficient—condenses hands-on processing time to as little as 30 minutes, images an array in less than 5 minutes, and operates unattended overnight 
  • Flexible—supports gene expression and genotyping studies on multi-format array plates 
  • Accurate—designed to deliver high-quality, consistent data by processing multiple samples under identical conditions 
  • Adaptable—creates flexible workflows and sample registration via the GeneChip Command Console (AGCC) software
Automated target preparation instruments for microarray analysis
Thermo Fisher Scientific has partnered with Hamilton Robotics to provide you with the latest in ultra-compact and precision-based liquid-handling instrumentation. The NIMBUS Target Preparation Instrument features high-end manufacturing and exceptional pipetting performance at an affordable price.  
Product specifications
GeneChip System 3000 GeneTitan MC Fast Scan Instrument
Dimensions
GeneChip Scanner 3000 with AutoLoader: 22.5 x 31 x 44.5 in. (57.2 x 78.7 x 113 cm)
GeneChip Fluidic Station 450: 28 x 16.1 x 15.8 in. (71.1 x 41 x 40.2 cm)
55” W x 33” D x 26” H (139.7 cm W x 83.82 cm D x 66 cm H)
Weight
GeneChip Scanner 3000 with AutoLoader: 105 lb (47.6 kg)
GeneChip Fluidic Station 450: 80 lb (36.3 kg)
147 kg
Ordering information
GeneChip System 3000
GeneTitan MC Instrument
NIMBUS Target Preparation Instrument
N° de catalogue Nom Taille
902169 Each
For Research Use Only. Not for use in diagnostic procedures.