GeneChip™ Fluidics Station 450
Applied Biosystems™

GeneChip™ Fluidics Station 450

The GeneChip Fluidics Station 450 is used for the wash and stain operation of GeneChip arrays.
Have Questions?
Catalog NumberQuantity
00-00791 each
Catalog number 00-0079
Price (USD)
-
Quantity:
1 each

GeneChip™ Fluidics Station 450
The GeneChip Fluidics Station 450 is used for the wash and stain operation of GeneChip arrays. The instrument includes design advancements to provide improved ease of use and true walk-away freedom dramatically improving efficiency in your genetic analysis.

The GeneChip Fluidics Station 450 is 110V and 220V compatible.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Dimensions (L x W x H)15 13/16 x 16 1/8 x 28 in. (40.2 x 41.0 x 71.1 cm)
For Use With (Application)GeneChip array processing
Product LineGeneChip
Quantity1 each
Weight (English)80 lb.
Unit SizeEach

Frequently asked questions (FAQs)

What is the part number for peristaltic tubing used inside the Fluidics Station 450 (Cat. No. 00-0079)?

The part number for Tubing, Peristatic Pump, 8.5" Long is 400110, which contains one tube per pack.

For the Fluidics Station 450 (Cat. No. 00-0079), how much wash buffer should I use for a single run when processing expression microarrays?

For most expression microarrays, the amount of wash buffer used during a single run is:
Wash A: approximately 150 mL
Wash B: approximately 50 mL

We recommend checking the user guide for your specific microarray for further details.

What fluidics protocol should I run for my Affymetrix array?

The array format and recommended fluidics protocols can be found in the packet insert of the array. This information can also be found in the the following document: Compatible Fluidics Script Table (https://assets.thermofisher.com/TFS-Assets/LSG/brochures/compatible_fluidics_script_table.pdf)
Note: Fluidics recommendations are based on the assumption that Affymetrix labeling kits and Hyb Wash Stain kits are being used.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What components are needed to run the Affymetrix Mouse Diversity Genotyping Array?

The components include: Affymetrix Mouse Diversity Genotyping Array
Affymetrix Genome-Wide Human SNP Nsp/Sty Assay kit 5.0/6.0
Affymetrix Power Tools (sed to run a modified BRLMM-P algorithm for genotype calling, GeneChip Fluidics Station 450
GeneChip Hybridization Oven 645
and GeneChip Scanner 3000 7G.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.

What is the minimum order size for the Mouse Diversity Genoytping array?

Customers must purchase a minimum of 60 arrays to begin a study, because this number of samples is required for BLRMM-P to cluster the data properly. For subsequent orders, the minimum order is approximately 30 arrays.

Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.