Search
CellTrace Violet Cell Proliferation Kit Protocol |
The CellTrace Violet (CTV) Cell Proliferation Kit is a powerful tool used for monitoring cell proliferation through dye dilution. In this CellTrace Violet protocol, live cells are covalently labeled with a very bright and stable dye which is retained within the cells and evenly distributed to daughter cells during cell division allowing researchers to track distinct generations of proliferating cells. This process, known as CTV staining, results in each cell generation appearing as a different peak on a flow cytometry histogram, facilitating precise evaluation of cell proliferation. These methods provide valuable insights into cellular behaviors and responses, making the CellTrace Violet kit a key tool for flow cytometric visualization of cell generations.
| CellTrace Violet | |
|---|---|
| Excitation/Emission (in nm) | 405/450 |
| Standard filter set | Pacific Blue |
| Storage conditions | ≤–20°C |
CellTrace Violet dye is a fluorescent dye used for tracking cell proliferation. It is commonly used in flow cytometry and microscopy to label live cells. The dye binds covalently to intracellular proteins, allowing researchers to monitor cell division and track cell populations over time. CellTrace Violet dye is known for its bright fluorescence and stability, making it suitable for long-term studies.
Yes. After labeling the cells with CellTrace Violet dye, you can fix them using aldehyde-based fixatives such as paraformaldehyde (PFA) or formaldehyde. This process helps to preserve the fluorescence and maintain the integrity of the labeled cells for subsequent analysis.
Cell Viability, Proliferation, and Cell Cycle Information
Find educational resources for monitoring cell function
Cell Proliferation Assay Protocols
Discover protocols for various applications to study cell proliferation.
Cell Cycle & Proliferation Pathways
Investigate signaling pathways and signal transduction.
Fluorescence SpectraViewer
Use SpectraViewer to easily compare excitation and emission spectra of fluorophores and reagents.
Cell Proliferation Assays for Flow Cytometry
Explore flow cytometry dyes, kits, and antibodies for cell proliferation.
Cell Proliferation Assays
Learn more about assays and reagents to evaluate cellular proliferation in flow cytometry, imaging, and microplate applications..
Microplate Assays for Cell Proliferation
Explore high-throughput screening of cell proliferation.
Cell Analysis Support Center
Find technical information, tips and tricks, and answers to everyday problems.
For Research Use Only. Not for use in diagnostic procedures.