Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Orbitrap / 5 Considerations When Selecting Your Next Research Mass Spec
5 Considerations When Selecting Your Next Research Mass Spec
By Scott M. Peterman, PhD, Sr. Product Marketing Manager, Life Science Mass Spectrometry, Thermo Fisher Scientific 01.04.2019
The Pursuit
Pursuit of high-throughput sample phenotyping through label-free quantitation (LFQ) using LC-MS requires substantial instrument performance to enable researchers to evaluate their experimental hypotheses. The goals of the assay are straightforward – profile as much of the proteome as possible with minimal sample preparation enabling you to determine specific expression profile differences based on donor and/or groups. This simply requires acquiring the data, processing it, and making sense of it using state-of-the-art liquid chromatographic separation techniques, mass spectrometers and bioinformatics pipelines.
Orbitrap Lumos
Thermo Scientific Orbitrap Fusion Lumos Tribrid Mass Spectrometer.
While that seems easy enough, especially given the continued advancements of today’s ‘state-of-the-art’ mass spectrometers and bioinformatics pipelines, the workflow still presents significant challenges. For example, profiling is defined as first confidently identifying and sequencing a peptide, and then being able to determine its response, generally in the form of integrated area-under-the-curve (AUC) when performing LC-MS. To perform untargeted profiling, the most successful methods are accomplished using high-resolution accurate mass (HRAM) MS for quantitative (and to a large degree qualitative) analysis coupled to MS/MS acquisition for qualitative sequencing analysis.
Thus, once the peptide sequence is matched, the chemical formula can be determined establishing the theoretical m/z values of the precursor isotopes at the measured charge state.
A mass tolerance window is established around the theoretical m/z values and extracted ion chromatograms are done per isotope, integrated AUC values determined and recorded, and the measured expression level for that peptide in a particular sample is known.
The width of the extraction mass tolerance is thus dependent on the performance of the mass spectrometer to reproducibly perform HRAM MS measurements and once determined by the researcher, the measured intensity is determined for each set of isotopes per identified peptide, integrated, and reported as the AUC. This value (normalized or not) is then compared to the measured AUC value for that peptide across all other samples to determine if it is “significant” or not (i.e., showing expression level changes based on biological condition).
The Caveat
To maximize the breadth and depth of proteome coverage, experimental parameters employ the shortest possible liquid chromatography gradients to manage throughput, resulting in very complex full scan mass spectra at any retention time point covering a very wide dynamic range. Therefore, the analytical capabilities of the mass spectrometer must do the following: scan really fast, maintain high intra-scan dynamic range, and acquire high-quality data for every spectrum and precursor.
Data quality can be defined by acquiring data with sufficient mass resolution and accuracy to completely separate measured signal between each precursor ion and maintaining very high mass measurement accuracy as well as product-ion rich MS/MS spectra. Successfully acquiring data in this manner enables the automated data processing software to confidently dig deep in the grass (or baseline) for additional peptides to achieve maximum proteome coverage.
The Assumption
Sadly, the assumption is that data acquisition speed is the primary factor governing experimental success. Time-of-flight (TOF) instruments certainly can acquire data extremely fast with new instruments reporting effective scan rates of 50 to 100 Hz and have high resolution (ca. 30,000 to 50,000) and accurate mass measurement (ppm) capabilities. In addition, state-of-the-art TOF mass spectrometers utilize ion mobility spectrometry to further improve selectivity prior to mass spectral analysis.
Complex biological samples, however, require significantly greater mass spectral performance to address the requirements listed above. The Thermo Scientific Orbitrap Fusion Lumos Tribrid mass spectrometer is unique in geometry and capabilities, maximizing the analytical performance required for LFQ with the top 5 reasons listed below:
  • The Orbitrap mass analyzer is capable of routinely acquiring mass spectra at a resolution setting of 120k, 240k and 500k (at m/z 200) as well as maintaining exceptional mass accuracy (< 3 ppm externally calibrated) for all precursors regardless of measured ion intensity and every spectrum
  • The dual pressure linear ion trap and ion routine multiple enables almost 100% duty cycle through parallel MS and MS/MS spectral acquisition, maintaining acquisition rates ca. 50 Hz
  • A high-resolution quadrupole mass analyzer capable of performing precursor isolation down to 0.4 Da while maintaining high transmission efficiency to further simplify resulting product ion spectra for increased accuracy in performing automated data processing
  • Automatic gain control and maximum ion filling — enables accurate quantitation across samples, instruments, and laboratories to enable method democratization
  • Extension of the Thermo Scientific FAIMS Pro interface to further enhance instrument performance by introducing orthogonal selectivity.
The combination of the unique features is ideal for maximizing LFQ experimental success. Utilizing parallelized HRAM MS and linear ion trap MS/MS acquisition provides researchers much higher resolution for MS acquisition in the Orbitrap mass analyzer while acquiring ca.50 full scan MS/MS spectra in the linear ion trap — all in one second. Thus one can maintain 7-10 MS data points across 10-second wide chromatographic peak widths AND still sample ca. 350-500 precursors with the highest degree of selectivity and sensitivity.
Since the precursor is measured with exceptionally high mass accuracy, it reduces the levels of false positives for the corresponding unit resolution product ion spectrum, resulting in extremely efficient processing speeds.
The Last Challenge
Last challenge of LFQ: automated data processing to determine expression levels of all peptides. Resolution and mass accuracy play a major role in the quality of the data output. Measuring and extracting the measured ion intensity for a specific peptide can be compromised by co-eluting isobaric peptides in the same or different charge states. Data acquired at lower resolution (50,000) can result in “shoulders” observed in the subsequent XIC peak in one sample while not in the others, altering the determined expression levels.
The second challenge is the stability of the mass measurement accuracy. When performing XIC analysis for an isotopic m/z value, an extracted mass tolerance is defined by the user to determine the measured ion intensity at that MS data point enabling the reconstruction of the XIC peak. If the stability of the mass measurement accuracy is not sufficiently high, wider windows (±10 to 30 ppm) are needed to eliminate dropping data points. The result is potentially compromised results requiring either manual interrogation, which is almost impossible given the amount of data within a sample, or extension of the study by acquiring technical replicates that extend the study to a point that is not really feasible.
As stated above, the Orbitrap Fusion Lumos MS acquires data at very high resolution and maintains exceptionally high mass measurement accuracy for more ions, spectra, and samples while maintaining effective precursor sampling rates as fast as most TOF instruments. Thus automated data processing becomes straightforward in determining isotopic patterns, accurate mass measurements, and accurate data extraction over a wide dynamic expression level range on complex samples.
Learn More
Top 5 reasons to upgrade to a Tribrid mass spectrometer white paper
Orbitrap Tribrid Mass Spectrometer
Grant writing support
Upgrade Smarter: Trade In Your Current LC-MS System and Boost Lab Performance Without Breaking Budget
Is your current mass spectrometer limiting your lab’s capa...
Read More
Redefining sensitivity and selectivity in steroid quantification
Advancing Steroid Quantification: Unlocking Sensitivity and Selectivity with the Thermo Scientific Stellar Mass Spectrometer
Quantifying steroids in biological matrices has always prese...
Read More
Sheffield teaching hospital NHS foundation
Beyond Blood: Sheffield Teaching Hospitals NHS Foundation Trust Develops HRAM-Powered Method for Post-Mortem Toxicology Analysis
In a recent study published in the Journal of Analytical Tox...
Read More
GLP-1 image
Five Key Advantages of the Thermo Scientific LC-MSn Solution for Ultra-Sensitive GLP-1 Quantitation
Bioanalytical analysis using LC-MSn requires precise, accura...
Read More
Scott M. Peterman, PhD
Scott Peterman, Ph.D., is Senior Product Marketing Manager for Life Science Mass Spectrometry of the Chromatography and Mass Spectrometry Division of Thermo Fisher Scientific.
Dioxin Testing Using GC MS MS Confidently Detecting the Hidden Dangers in the Food Chain
Top Six Reasons Upgrading Your Small Molecule Structural Elucidation Approach May Be in Order