Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Columns and Consumables / Adding μPAC HPLC Columns to Your Portfolio
Adding μPAC HPLC Columns to Your Portfolio
By AnalyteGuru Staff, 02.25.2022
Bioresearch 022522.jpg
For many years Thermo Fisher Scientific has worked closely with PharmaFluidics, inventor and manufacturer of micro pillar array columns (μPAC) for low-flow liquid chromatography (LC) omics experiments. This partnership has generated exciting results. For example this poster from HUPO and journal article demonstrate the high-quality, reproducible chromatography performance you can expect when our products are used together.
In 2021 the PharmaFluidics team joined Thermo Fisher Scientific. The merger resulted in the formal release of Thermo Scientific™ μPAC™ HPLC Columns in January 2022, an exciting step in our journey.
Pharmafluidics 800 022522.jpgμPAC HPLC columns are built by precisely micromachining chromatographic separation beds into silicon wafers. This approach produces columns with perfect order, providing the following chromatographic characteristics:
Sample coverage– µPAC HPLC columns provide comprehensive coverage that is ideal for proteomics, metabolomics, and lipidomics. When used for single-shot proteome analysis, nearly 10,000 identifications can be achieved so you can discover more about your sample.
Column-to-column reproducibility– Each column is manufactured using the same lithographic mask, providing consistency and performance from column-to-column for a high level of confidence in your data.
Flow rate flexibility– µPAC HPLC columns can be operated at moderate LC pump pressures up to 350 bar over a wide range of flow rates (100 – 1,000 nL/min) for method flexibility.
The newly expanded Thermo Scientific low-flow column range offers multiple column formats and column chemistries to cover all your bottom-up and top-down experiments, and budgetary needs. More information on our range can be found here.
Welcoming new colleagues and customers to Thermo Fisher Scientific has been an amazing start to the year. I’m looking forward to seeing what the future holds as creative minds come together as one team to service research scientists in new ways.
Welcome to the family, μPAC HPLC Columns!
Discover more.
Excedion Pro Banner
Discover. Innovate. Exceed. — Introducing the Thermo Scientific Orbitrap Excedion Pro Mass Spectrometer
Mass spectrometry is entering a new era of precision, perfor...
Read More
Thermo Fisher Wins Lab Innovation Award for New Stellar Mass Spectrometer
Thermo Fisher Wins Award for Stellar Mass Spectrometer
Celebrating innovation in mass spectrometry We are delighted...
Read More
AAV-SEC Analysis: Top 3 Takeaways From the Latest Article
AAV-SEC Analysis: Top 3 Takeaways From the Latest Article
What is the best column for separating AAV size variants? Th...
Read More
Unveiling the Power of the Stellar Mass Spectrometer in Metabolomics
Unveiling the Power of the Stellar Mass Spectrometer in Metabolomics
In the intricate world of metabolomics, understanding the co...
Read More
AnalyteGuru Staff
Don’t be Caught Off Guard by Ever-Changing Nitrosamine Regulatory Landscapes
HPLC Troubleshooting: Why Signal-to-Noise Ratio Determines Limit of Detection