Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Oligonucleotide / Consider Semi-preparative Columns to Improve Your Oligonucleotide Analysis
Consider Semi-preparative Columns to Improve Your Oligonucleotide Analysis
By Gilles Jaouen, LC Columns Product Management, Thermo Fisher Scientific 07.30.2024
The field of oligonucleotide therapeutics is expanding due to the push toward personalized medicine and continued research into the development of therapies for rare diseases. However, oligonucleotides are difficult to produce in high yields with high purity and formulate for efficient delivery and stability. Are you faced with the challenge of effectively keeping up with the growing demand for oligonucleotide testing? Are you seeking a way to accurately separate and purify complex oligonucleotides samples? Here are two questions that will help you consider a new column format that is reliable and cost-effective:
How can I carry out oligonucleotide analysis in high yields and be cost-effective?
Your laboratory can experience easy integration into your existing setup and smooth scalability from analytical to semi-preparative scale at high throughput, without sacrificing consistent and reliable performance. The industry leading Thermo Scientific DNAPac RP HPLC Columns are now available in a semi-preparative format for oligonucleotide purification accessible on a semi-preparative LC system. The unique performance of the DNAPac RP microporous structure achieves efficient separation at a preparative scale, even with instruments operating up to 10 mL/min.
How would a semi-prep column format improve my oligonucleotide analysis?
The unique chemistry of the DNAPac RP semi-preparative column delivers unmatched performance across a wide range of pH, temperature, and mobile phase compositions, enabling precise separations and accurate quantification, regardless of your experimental conditions. Specifically designed for ion-pair reversed-phase (IP-RP) separations, the column’s polymeric backbone bead provides sharper peaks and less band broadening, which are often seen with silica columns. The column’s wide operating pH range (0-14) and high-temperature range (up to 110°C) make it the ideal support for method development.
The DNAPac RP column chemistry is based on a unique super-macroporous structure with a pore size range of up to 2000 Å, offering excellent separation of both short and long single- and double-stranded nucleic acids, including RNA modalities such as intact mRNA.
Its biocompatible inert stainless-steel hardware reduces undesired secondary interactions, increasing recovery and ensuring accurate and reproducible results. By reducing unwanted interactions, the column preserves the integrity of your samples and maintains the purity of your analytes.
To learn more about the DNAPac RP semi-preparative columns, check out these online resources:
  • Read the flyer: Revolutionize your oligonucleotide analysis with game-changing DNAPac RP columns
  • View the product page: DNAPac™ RP Columns
Visit our LinkedIn page #oligonucleotides #DNAPac
Hand placing a missing piece into a white jigsaw puzzle.
Blueprint for QC Ready mRNA Mapping
mRNA therapeutics demand analytical methods that are fast, r...
Read More
Innovation in Biopharma analysis image
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Biopharmaceutical pipelines are expanding rapidly, with incr...
Read More
freedom to fragment
Collision-Based Ion-activation and Dissociation
Introduced nearly 80 years ago, collision-based ion-activati...
Read More
AAV-SEC Analysis: Top 3 Takeaways From the Latest Article
AAV-SEC Analysis: Top 3 Takeaways From the Latest Article
What is the best column for separating AAV size variants? Th...
Read More
Gilles Jaouen
Gilles Jaouen completed his Chemical Engineering Msc at Lille National Engineering School of Chemistry and MBA at Insead (France). Following graduation, he contributed to several market-leading groups on developing, marketing and managing chromatography columns and consumables portfolio. In 2022, he joined Thermo Fisher Scientific, where he currently leads the LC Columns Product Management team, including BioLC and DNA Pac column family.
Size Exclusion Chromatography of Adeno-associated Viruses
Why Everyone is Talking About PFAS and EPA Method 1621