Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Liquid Chromatography / HPLC Method Transfer Simplified: How to Save Your Time and Sanity
HPLC Method Transfer Simplified: How to Save Your Time and Sanity
By Dr. Kimberly M. Lincoln, HPLC, Thermo Fisher Scientific 08.25.2021
Do you struggle to transfer validated HPLC methods between labs or different instruments? Is your business losing money or missing critical deadlines due to the struggle? You are not alone.
Transferring a validated method from one HPLC system to another is rarely a smooth or enjoyable process. Differences in the hardware designs between two instruments can affect chromatographic results. You might tinker with parameters to replicate the method or even change the tubing or mixers to match the dead volume, which may result in re-qualification of the system — a lot of extra time and mental agony.
The good news is your HPLC method transfer process does not have to be complicated. With intelligent tools like a tunable gradient delay volume (GDV) built into the Thermo Scientific™ Vanquish™ Core HPLC system, you can simplify and expedite method transfer hassle-free.
Here is a full breakdown of the hardware and software features within the Vanquish Core HPLC system, making adopting outside methods simple.
How a tunable GDV simplifies method transfer
For gradient HPLC methods, the instrument GDV plays a central role in successful method transfer. Defined as the volume from the point of mixing of the eluents to the column head, modifications in the GDV between instruments can cause a loss of retention time precision and resolution.
A tunable GDV can help conserve analyte elution profiles, empowering you to transfer methods easier and faster. The autosampler in the Vanquish Core HPLC system features an integrated metering device as part of the flow path. This feature enables you to tune the GDV of the system from 0–230 µL.
If you need an adjustment volume greater than 230 µL or want the flexibility of a larger GDV, you can opt for the method transfer kit. This optional kit allows you to increase the GDV up to 430 µL without altering the qualification status of your instrument. You can see a visual demonstration of how adjusting the GDV works here.
Full Vanquish Core HPLC system features to simplify HPLC method transfer
The Vanquish Core HPLC system is equipped with a full suite of hardware and software, to streamline your method transfer process:
  • Tunable GDV from 0 – 430 µL to match retention time and resolution.
  • Two unique column thermostatting modes to conserve analyte selectivity.
  • Custom injection programs for strong solvent samples to minimize peak distortion from solvent mismatches.
  • Simple setup and use with Thermo Scientific Chromeleon CDS.
  • Full regulatory compliance by audit trail tracking and qualification.
These additional instrument solutions give you the flexibility to transfer methods between labs or another vendor system efficiently.
No more pulling your hair out over missed deadlines and lost revenue.
Productivity and confidence will define you.
Save cash money by trading in your HPLC system today
Are you ready to stop wasting time and money on failed method transfers?
For a limited time, you can save big on your new Vanquish Core HPLC system by trading in your archaic money pit. Before you do so, make sure to take our virtual 3D product tour to get a detailed look at the system features and benefits.
Once you commit to making your life better, you can request a quote online in minutes using our digital instrument builder.
Your future is bright, so let the world leader in serving science be your light.
Amino acid chromatogram showing labeled peaks over a 7-minute separation time.
How to Cut HPLC Sample Preparation Time for Derivatizing Amino Acids
Derivatizing amino acids doesn’t have to take hours to do....
Read More
Elevate Your HPLC Performance with a Modern CDS
Elevate Your HPLC Performance with a Modern CDS
You may have the best HPLC system on the market, but to maxi...
Read More
Vanquish CAD P Series blog cover image
Episode 4: Introducing a More Confident Era in HPLC-CAD Analysis
The first Charged Aerosol Detector (launched in 2005) set ne...
Read More
The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
Episode 1: The Benefits of Charged Aerosol Detection (CAD) in High-Performance Liquid Chromatography (HPLC)
High-Performance Liquid Chromatography (HPLC) is a cornersto...
Read More
Dr. Kimberly M. Lincoln
Dr. Kimberly M. Lincoln is a marketing specialist in the HPLC division at Thermo Fisher Scientific. She holds a Ph.D. in bioinorganic chemistry and a B.S. in chemistry, with professional experience in digital marketing, medical device manufacturing, and chemical synthesis. Kimberly enjoys writing educational chemical content as a guide to help customers solve analytical problems. She is passionate about living a clean lifestyle and keeping the planet healthy for future generations. In her spare time, Kimberly loves to cook, cycle, attend rock concerts, and spend time with her pit bulls.
Letting Go Has Its Rewards Consider Replacing Your Old IC System
Nitrosamine Impurity Analysis: Every Tool You Will Ever Need