Thermo Fisher Scientific
Your educational resource for biopharma, pharma, environmental, food and agriculture, industrial, and clinical labs
  • Categories
    • Advancing Materials
    • Advancing Mining
    • AnalyteGuru
    • Analyzing Metals
    • Ask a Scientist
    • Behind the Bench
    • Biotech at Scale
    • Clinical Conversations
    • Examining Food
    • Identifying Threats
    • Illuminating Semiconductors
    • Life in Atomic Resolution
    • Life in the Lab
    • OEMpowered
    • The Connected Lab
  • About Us
  • Contact
Accelerating ScienceAnalyteGuru / Biopharmaceutical / Unveiling Hidden Impurities: LC-HRMS for Biopharmaceutical Analysis
Unveiling Hidden Impurities: LC-HRMS for Biopharmaceutical Analysis
By Lizzie Gallagher, Market Development Manager, Pharma and BioPharma, Thermo Fisher Scientific 03.26.2024
This is the fifth in a five-part BioPharma Blog Series.
Have you ever wondered how advanced scientific techniques can unveil hidden impurities in the biopharmaceutical world?
Liquid chromatography high-resolution mass spectrometry (LC-HRMS) is one such cutting-edge method gaining traction in untargeted screening and identification of small-molecule impurities, particularly leachables from container closure systems.
In contrast to controlled extractables studies, leachables studies are carried out on the final drug formulation, where the presence of the protein drug matrix, containing relatively high levels of the active protein(s) and excipients, presents an analytical challenge for LC-HRMS analysis. Due to this complexity, confident proposal of unknown compound structures typically requires high-resolution mass spectra with MS2 and MSn fragmentation.
At a recent USA based conference, a poster was presented that detailed how the Thermo Scientific Vanquish Horizon UHPLC system coupled to a Thermo Scientific Orbitrap Exploris 240 MS compared to a typical LC-QTOF screening method, and the results are available for you to view.
Download our free impurities poster now.
Related information
Blog 1: Revolutionizing Host Cell Protein Detection and mAb Analysis in Biopharmaceuticals
Blog 2: Poster Series: A Journey into the World of Biopharma Column Chemistry
Blog 3: Take a Step Closer to Unlocking Full Potential of AAV Gene Therapy – Download the Poster
Blog 4: Unlocking the Future of Medicine: Exploring Therapeutic Oligonucleotides
Download our conference posters to see how we applied advanced analytical technology to achieve the best results for oligonucleotide analysis
On LinkedIn? Visit our LinkedIn page #biopharmaceutical #UHPLC
Innovation in Biopharma analysis image
Scaling Up Biopharmaceutical Characterization with the Orbitrap Ascend BioPharma Tribrid Mass Spectrometer
Biopharmaceutical pipelines are expanding rapidly, with incr...
Read More
laptop with ThermoFisher logo
Understanding Nitrosamine Drug Substance-Related Impurities (NDSRIs)
Introduction Nitrosamine drug substance-related impurities (...
Read More
5 Key Takeaways from LC Columns Tips and Tricks Webinar
The webinar, hosted by Hai Ning, Regional Marketing Manager ...
Read More
Advancing PFAS Detection in Pharmaceutical Manufacturing with LC-HRAM-MS
Advancing PFAS Detection in Pharmaceutical Manufacturing with LC-HRAM-MS
In the realm of pharmaceutical manufacturing, the detection ...
Read More
Lizzie Gallagher
Lizzie Gallagher is a market development manager for the pharma and biopharma market at Thermo Fisher Science, having spent a number of years in both industry, and sales and marketing. Lizzie has widespread experience with the challenges and trends of the biopharma and pharma market.
Practical Analysis Solutions to Enhance Water Quality for a Safer Future
Advanced LC-MS Tools and Software Solutions to Ease Your Metabolite Identification Challenges