Applied Biosystems SeqStudio Genetic Analyzer for Human Identification (HID) is Now Available
This 4-capillary fragment analysis platform brings trusted technology and Scientific Working Group on DNA Analysis Methods (SWGDAM) validated workflow to forensic DNA laboratories.
The Applied Biosystems SeqStudio Genetic Analyzer for HID is available to support small to medium sized forensic DNA analysis and paternity laboratories who need a lower throughput (four capillary) CE system that is affordable and easy to use.
The SeqStudio Genetic Analyzer for HID utilizes a unique integrated cartridge that combines the capillary array, polymer reservoir and anode buffer, which simplifies instrument set-up and minimizes hands-on processing time. Watch the video to see how easy it is to set up and use the SeqStudio system.
The system has been validated according to SWGDAM guidelines across a broad range of Applied Biosystems™ STR Amplification Kits. Forensic analysts can obtain STR-based DNA profiles comparable to those generated on the gold-standard 3500 Genetic Analyzer. Optimized data collection software provides reduced profile editing and seamless integration with GeneMapper™ ID-X Software v 1.6. The Security, Audit and E-signature (SAE) software module provides the protection and traceability needed by forensic DNA and paternity laboratories.
For more information about the SeqStudio Genetic Analyzer for Human Identification, please visit thermofisher.com/hid-seqstudio
For Research, Forensic or Paternity Use Only. Not for use in diagnostic procedures.