Building a Better Sample QC Workflow: From Extraction to Sequencing
Article Summary
Successful genomics experiments don’t begin with sequencing—they begin with sample quality.
Every downstream application, from PCR to next-generation sequencing (NGS), depends on starting with nucleic acids that are accurately quantified and free from contaminants that could compromise results. Establishing a consistent quality control workflow helps laboratories improve reproducibility while reducing delays caused by failed samples.
Consistent quality control workflows
Step 1: Start with High-Quality Extraction
Quality control begins during sample preparation. Consistent extraction methods help minimize contaminants while maximizing nucleic acid recovery. Even well-established protocols can produce variable results depending on sample type, operator technique, or reagent quality, making post-extraction QC an essential checkpoint.
Step 2: Evaluate Concentration and Purity
Following extraction, researchers should verify both concentration and sample purity before investing additional time and resources. Microvolume UV-Vis spectrophotometry allows laboratories to rapidly measure nucleic acid concentration while simultaneously assessing purity ratios and reviewing full absorbance spectra. This immediate feedback helps identify samples that may require additional cleanup before moving forward.
Step 3: Match QC to the Application
Not every experiment demands the same level of quality control. Routine molecular biology applications may require only concentration and purity verification, while highly sensitive workflows—such as NGS library preparation—often incorporate fluorescence-based quantification to improve measurement specificity. Selecting QC methods based on downstream requirements helps laboratories balance speed with confidence.
Step 4: Standardize the Workflow
One of the simplest ways to improve reproducibility is to standardize sample QC across operators and projects. Consistent blanking procedures, cleaning protocols, measurement techniques, and acceptance criteria reduce variability and make it easier to compare results over time. Documented QC practices also support training, quality systems, and collaborative research environments.
Conclusion: Turning QC into Better Results
Quality control should not be viewed as an additional task—it is an investment in experimental success.
A structured workflow that combines reliable extraction, rapid purity assessment, appropriate quantification, and standardized laboratory practices helps researchers reduce repeat experiments, improve data quality, and make better-informed decisions before every sequencing run.
As genomics research continues to advance, strong quality control remains one of the simplest and most effective ways to improve confidence in every result.
Frequently Asked Questions
  • Sample quality influences the reliability of downstream applications. Verifying nucleic acid concentration and assessing sample purity before PCR, qPCR, or NGS can help improve reproducibility and reduce the likelihood of repeat experiments.
  • A typical nucleic acid quality control workflow includes consistent sample extraction, measurement of concentration and purity, selection of application-appropriate quantification methods, and standardized laboratory procedures for evaluating sample quality before downstream analysis.
  • Concentration measurements determine how much nucleic acid is present, while purity assessments help identify contaminants that may interfere with downstream applications. Evaluating both provides a more complete understanding of sample quality.
  • Microvolume UV-Vis spectrophotometry enables rapid measurement of nucleic acid concentration while simultaneously providing purity ratios and the full absorbance spectrum to help evaluate sample quality after extraction.
  • Fluorescence-based quantification is commonly used when greater measurement sensitivity or specificity is required, such as before next-generation sequencing (NGS) library preparation or when working with low-concentration samples.
  • Many laboratories use absorbance to quickly assess concentration and purity following extraction, then use fluorescence-based quantification when greater sensitivity or application-specific accuracy is needed before downstream workflows.
  • Standardizing blanking procedures, cleaning protocols, measurement techniques, and acceptance criteria can reduce variability between operators, improve consistency across projects, and make results easier to compare over time.
  • Sample quality can be influenced by extraction methods, sample type, reagent quality, operator technique, and the presence of contaminants that may affect downstream enzymatic reactions or sequencing workflows.
  • Evaluating sample quality at multiple stages of the workflow can help identify low-quality or contaminated samples before downstream applications, reducing troubleshooting, repeat experiments, and delays.
  • Consider the requirements of your downstream application, the appropriate methods for measuring concentration and purity, laboratory standardization practices, and acceptance criteria that support consistent, reproducible results.
Kyle Mcnally, Thermo Fisher Scientific
Written by:
Kyle Mcnally
Senior Market Development Specialist – UVN , Thermo Fisher Scientific
Kyle Mcnally works with the UV-Vis spectroscopy team to support global marketing campaigns.
Read more Mcnally, Kyle