Designing for Predictable Scale-Up in Pharmaceutical Extrusion 
Overview 
Predictable scale-up in pharmaceutical extrusion depends on preserving the material’s experience inside the extruder. The thermal load, mechanical energy input, and residence time are key, as opposed to simply increasing equipment size or throughput. As extruder diameter increases, heat transfer dynamics, energy distribution, and residence time profiles change in non-linear ways. These changes can subtly affect dispersion quality, amorphous stability, milling performance, and ultimately critical quality attributes such as dissolution and tablet strength. Geometric similarity between lab and production equipment enables meaningful parameter-based scale-up, allowing teams to align shear, temperature, and exposure time across scales. By considering these factors early in development and designing processes that preserve key controls, pharma manufacturers can reduce variability, maintain consistent product performance, and make scale transitions more predictable and robust. 
Designing for predictable scale-up in pharmaceutical extrusion 
Why heat transfer, mechanical energy, and residence time deserve attention early in development 
Scaling a pharmaceutical process rarely feels dramatic at first. In most cases, it appears to be a logical progression: a larger barrel diameter, increased throughput, or a transition to a different facility. From an operational perspective, the path seems straightforward. 
However, teams that have navigated late-stage scale transitions know that process behavior does not always change proportionally with equipment size. A formulation that performs reliably at laboratory scale can exhibit subtle but meaningful shifts at pilot or production scale. Dissolution profiles of the final product may change slightly. Thermal process margins may narrow. Variability can increase in ways that are difficult to attribute to a single adjustment. 
These outcomes are not typically the result of poor engineering. More often, they reflect the underlying physics of scaling. 
In recent industry discussions, including our contribution to The Medicine Maker on continuous manufacturing and drug development, a consistent theme has emerged: scale decisions are increasingly being considered earlier in development. This shift reflects a growing recognition that scaling influences formulation behavior long before formal technology transfer begins. 
To understand why this matters, it is useful to examine what changes when equipment size changes—and what must be preserved. 
What changes when diameter changes 
When transitioning from laboratory to pilot or production extrusion, three physical factors must be carefully aligned across scales: thermal load, mechanical energy input, and residence time profile. 
To ensure a final product behaves similarly at small and large scale, the material must undergo the same thermal stress, experience the same mechanical energy per unit mass, and be exposed to comparable residence time distributions. 
Heat transfer and thermal design 
As screw diameter increases, surface area increases with the square of the diameter, while material volume increases with the cube. The ratio between available heat-transfer surface and process volume therefore shifts. Larger systems inherently have less surface area per unit volume to remove or supply heat. 
This does not mean larger systems are unstable. It means thermal management must be designed appropriately. 
In well-designed small-scale processes, the system should approach adiabatic behavior once stable operation is reached. External barrel heating is appropriate during start-up, but once the process is steady, the dominant energy input should come from the mechanical work of the screws. If small-scale development relies heavily on external heating or cooling, scale-up can become problematic later when the larger system does not have sufficient surface area to compensate. 
In practice, poor thermal alignment does not usually present as a dramatic failure. Instead, the process window may narrow, melt temperature may become more sensitive to minor fluctuations, and the final product may show increased variability. 
Thermal behavior changes with geometry. Predictable scale requires anticipating that relationship. 
Mechanical energy and specific mechanical energy (SME) 
Extrusion performance is not governed by temperature alone. The mechanical work introduced into the material—typically expressed as specific mechanical energy (SME or SMEC)—is central to material transformation. 
Changes in screw diameter, torque response, and fill behavior can alter the energy input per kilogram of material. Even if rotational speed appears proportionally scaled, the actual mechanical energy delivered to the formulation may differ. 
If SME differs between scales, the internal structure of the dispersion can shift accordingly. Because dispersion quality, amorphous stability, and melt homogeneity are directly linked to mechanical and thermal stress inside the extruder, deviations in SME can influence critical quality attributes (CQAs) of the final product. 
Maintaining comparable mechanical energy input across scales is therefore not optional—it is essential for preserving product performance. 
Residence time shapes exposure 
Residence time distribution (RTD) defines how long the material is exposed to thermal and mechanical stress. Larger extruders may exhibit different fill levels and altered residence time profiles if parameters are not adjusted deliberately. 
Segmented screw architecture allows control of conveying and kneading zones. However, this control must be translated proportionally across scales to maintain comparable exposure conditions. 
Heat, energy, and time together define the material’s experience inside the extruder. For scale-up to be predictable, that experience must remain comparable across sizes. 
Why this matters beyond the extruder 
Scale-up effects do not stop at the die. 
Research on downstream processing of hot-melt extruded amorphous dispersions has demonstrated clear relationships between upstream process conditions and downstream performance. Changes in strand structure influence milling efficiency. Milling influences particle size distribution. Particle size distribution affects tablet tensile strength and dissolution behavior. 
When thermal stress, mechanical energy, or residence time differ between scales, those differences propagate into measurable changes in CQAs. 
Scale-up challenges often emerge not because the process is flawed, but because the material does not “see” the same stress conditions at different scales. 
Geometric similarity enables parameter-based scale-up 
There are not competing scale-up approaches divided into “geometry-dependent” and “parameter-driven” systems. In extrusion, geometric similarity between equipment sizes enables meaningful parameter adaptation. 
When extruders are geometrically similar—maintaining proportional L/D ratios, screw design philosophy, and overall architecture—process parameters can be adjusted to recreate equivalent thermal, mechanical, and time stress conditions across scales. 
Design-of-experiments approaches can then be used to define overlapping design spaces based on residence time, melt temperature, and specific mechanical energy. Within those overlapping regions, scale-up becomes significantly more predictable. 
If geometric similarity is absent—for example, when transferring between fundamentally different machine designs or vendors—predicting behavior becomes more complex because the relationship between geometry and parameters changes. 
Geometric similarity does not eliminate the need for parameter control. It makes parameter-based scale-up achievable. 
Designing for continuity 
When scale is considered early, development discussions shift from equipment size to material experience. 
Questions become more focused: 
  • Can specific mechanical energy be matched across scales? 
  • Does the larger system have sufficient surface area to dissipate process heat? 
  • Is residence time distribution measured and aligned? 
  • Are screw geometries proportionally scaled? 
  • How will upstream process conditions influence downstream CQAs? 
These questions do not complicate development. They clarify it. 
In our recent discussion in European Pharmaceutical Manufacturer, we explored how preserving shear, temperature, and residence time across laboratory and production systems supports more predictable transfer. The same principle applies here: continuity in governing parameters is what stabilizes scale transitions. 
A formulation that behaves consistently across scales rarely does so by chance. It reflects deliberate preservation of thermal load, mechanical energy, and residence time conditions within geometrically comparable systems. 
A useful principle is this: 
Preserve the material’s experience across scales. 
When geometric similarity supports parameter alignment, scale-up becomes less about increasing size and more about maintaining continuity. 
That is where predictability begins. 
Frequently Asked Questions 
  • How early in development should scale considerations be incorporated into pharmaceutical manufacturing projects? 
    • Scale considerations should begin during laboratory-stage development, not at the point of formal technology transfer. Scale influences formulation behavior long before production equipment is introduced 
  • What are the key parameters to consider when using extrusion for pharmaceutical scale up? 
    • The key parameters depend on the active pharmaceutical ingredient’s response to physical factors introduced during extrusion: thermal load, mechanical energy input, and residence time profile. Reliance on external heat sources, the amount of specific mechanical energy delivered to the materials at various points in the system, and how the material is affected by thermal or mechanical stress are all important factors. 
  • What role does the geometric similarity of equipment play in enabling predictable scale-up? 
    • While a larger extruder may look like a scaled up smaller extruder, geometric similarity does not necessarily lead to identical processing. Care must be taken to maintain proportional length-to-diameter (L/D) ratios, screw design philosophy, and overall architecture. These make parameter-based scale-up achievable. When equipment sizes are geometrically aligned, process parameters such as melt temperature, residence time, and specific mechanical energy can be adjusted to recreate equivalent material stress conditions across scales. Geometric similarity does not eliminate the need for careful parameter control, but it makes meaningful parameter alignment possible. 
Dirk Leister
Written by:
Dirk Leister
Manager Applications & Customer Support , Thermo Fisher Scientific
Dirk Leister is a marketing manager at Thermo Fisher Scientific focused on extruder and compounder technology used in the pharmaceutical, food, and polymer industries.
Read more Leister, Dirk