Have you ever tried pipetting a viscous solution, only to find that your pipette could not accurately draw up the solution? Have you worried about damaging fragile cells or unstable molecular complexes when pipetting?
If you are working with samples that are difficult to pipette or can be damaged by pipetting, you should consider using wide bore pipette tips.
Wide bore pipette tips
Wide bore pipette tips are equipped with a distal end orifice that is nearly 70% larger than that of a standard pipette tip. These tips provide the added flexibility required for handling difficult-to-pipette samples.
Thermo Scientific™ ART™ Wide Bore Pipette Tips seal when exposed to potential contaminants, trapping them inside the barrier. Wide bore pipette tips are available in different volumes (200–1,000 mL) and packaging options (bulk, racked, nonsterile, sterile, non-filtered, and filtered).
Wide bore pipette tips are especially useful for the following applications:
Genomic DNA (gDNA)
Pipetting alone can shear gDNA and should be considered when considering a protocol that will include the transfer of gDNA. A wide bore pipette tip will keep the gDNA more intact and lessen the chance of shearing during pipetting.
Cellular materials
When working with fragile cellular samples such as macrophages, hybridomas, or hepatocytes, cellular damage is a big concern. Using a pipette tip that will not damage cells passing through the distal end should be considered in order to protect the cells during transfer.
Viscous samples
For those working with viscous solutions, the wider opening allows thicker, more viscous liquids to pass through by lowering the pressure needed for movement through the orifice and into the shaft of the tip. In general, by lowering the pressure needed to pass the sample through the orifice, the resistance of flow is decreased, and less stress is put on the sample during liquid handling.
Particulate samples
Using a wide bore pipette tip for samples that have particles (e.g., environmental/food-derived samples, beads, etc.) can prevent blockage in the distal end of the tip during pipetting. The user can save time, repeat pipetting, and prevent contamination of sample or pipette shaft with use of a wide bore pipette tip.
Additional considerations when using wide bore tips
It is always important to check the performance of your pipette and tips prior to use so that any adjustments can be made to obtain the best accuracy and precision. Also, there are different pipetting techniques that can be performed to better handle difficult-to-pipette samples. Please refer to the fundamentals of pipetting practices online guide for a comprehensive overview of pipetting techniques to improve your pipetting in the lab.
To find out more about Thermo Scientific ART Pipette Tips, visit thermofisher.com/art.
Medical supplies are something I have not been doing for years, and I must say you have got all the facts right about pipettes. Wide bore pipette is more commonly used, and I have been continuously supplying them to test labs across the state. The article is surely a great insight into different types of pipettes.
I was work as a lab Technician for more than 15 years in craftsman storage.I never knew about this fact about Pipette. thank you for sharing
very useful article for me, thanks
Everyone somehow knows about pipette, but I’m sure most don’t know this fact. It’s really great to get information here. Thanks for sharing.
Everyone somehow know what is a pipette? But I’m damm sure a lot don’t know this information. Thanks for sharing
Really some sort of unknown information. Thanks for sharing
REL is the noticeable producer of Mining Drilling Rigs
Do pipette has these difference really???
Setting a minimum goal for doors knocked a day is a great starting point. Tracking the outcomes of those door knocks are equally important because you want to know; 1) How many knocks it takes to get a good lead, 2) How many leads it takes to make a sell, 3) The areas you’ve already knocked, 4) Collecting customer info to covert more to sales.
Thank you for sharing a information about this pipette
what’s the Cost of this pipette?. Is it available in India
What is the maximum temperature that pipette withstand?
The Pipette is made on glass or plastic?