Preparing for Windows 11: An upgrade guide for your laboratory instrument
Two scientists in an unreal environment looking at a computer that is getting a Windows 11 Upgrade
Replacing an instrument workstation involves more than swapping out a PC. From software compatibility and data backup to instrument configuration and performance verification, thoughtful planning can help minimize downtime and support a smoother transition.
An instrument workstation is more than just a PC
Every laboratory eventually reaches the point where an instrument workstation needs to be replaced or upgraded; sometimes the catalyst is aging hardware, sometimes it’s a software upgrade. Increasingly, organizations are replacing Microsoft™ Windows™ 10 workstations as part of their transition to Windows 11.
Whatever the reason, it’s easy to underestimate what an instrument workstation actually represents.
Unlike a standard office computer, an instrument workstation is an integral part of the analytical system it supports. It hosts instrument control software, manages communication between the instrument and the operating system, stores configuration settings and calibration files, and often serves as the point where laboratory data begins its journey through downstream systems.
Replacing that workstation is therefore not simply an IT refresh; it’s a change to the environment that enables the instrument to function reliably, making thoughtful planning essential before any work begins.
Contact technical support to get started with your Windows 11 transition.
One of the biggest factors influencing a successful workstation transition is planning.
Organizations that experience the fewest disruptions typically begin preparing well before the replacement takes place. They identify which systems are affected, schedule downtime around laboratory operations, coordinate responsibilities between laboratory staff and IT, and ensure critical information has been backed up before work begins.
A successful transition often includes activities such as:
None of these activities is particularly difficult on its own. However, overlooking even one can extend downtime or delay the return of an instrument to service.
One of the most common misconceptions is that once Windows is installed, the project is complete. In reality, the operating system is only the foundation. Before an instrument can return to productive use, the workstation must be configured correctly, instrument software installed, communication with the instrument verified, calibration information restored, and system performance confirmed.
This distinction is important because laboratory productivity depends on the instrument—not the computer.
The objective isn’t simply to replace hardware. It’s to restore the complete instrument workstation environment so scientists can resume their work with confidence.
Why Windows 11 makes this more urgent
Microsoft’s end of support for Windows 10 has accelerated workstation replacement planning across many organizations. While Extended Security Updates may provide temporary relief for some environments, they do not eliminate the need to modernize aging instrument workstations.
For many laboratories, the Windows 11 transition provides an opportunity to replace older hardware, move to supported software configurations, and improve long-term supportability.
However, Windows 11 also brings additional planning considerations. Instrument software compatibility should be confirmed, supported workstation configurations identified, and replacement schedules coordinated with laboratory operations. Organizations should also understand which activities remain internal responsibilities—such as data backup, network configuration, and validation—and which activities may benefit from manufacturer support.
How Thermo Fisher Scientific can help
Transitioning an instrument workstation requires both IT coordination and instrument expertise.
While most internal IT organizations are well equipped to deploy Windows workstations, restoring an analytical instrument to an operational state requires familiarity with the instrument’s software, configuration, calibration, and performance verification.
  • Reduce instrument downtime – A structured workstation transition helps minimize disruption and supports a faster return to productive operation.
  • Preserve instrument performance – Calibration transfer, instrument calibration, and performance verification help ensure supported instruments are operating as expected following workstation replacement.
  • Reduce the burden on your team – By performing instrument-specific installation activities, Thermo Fisher helps reduce the workload placed on laboratory personnel and internal IT teams.
  • Factory-approved configuration – Supported Windows 11 workstations are configured using Thermo Fisher-approved hardware and software configurations.
Thermo Fisher’s Instrument Workstation Transition Service is designed specifically to support this phase of the project. For supported instruments, our field service engineers install a factory-approved Windows 11 workstation, reinstall and configure Thermo Fisher instrument software, transfer calibration files, perform instrument calibration, and verify successful operation through a standard injection test before returning the instrument to service.
Replacing an instrument workstation is an event every laboratory will face. Whether the driver is aging hardware, software modernization, or the transition to Windows 11, approaching the project with a structured plan can help reduce downtime, improve coordination, and restore instrument operation more efficiently.
By understanding the work involved, defining responsibilities early, and engaging the right expertise at the right time, laboratories can turn what might otherwise be a disruptive project into a predictable and well-managed transition.
If your organization is planning to replace Windows 10 instrument workstations, your Thermo Fisher representative or our technical support team can help you understand the transition process, clarify responsibilities, and schedule installation for supported systems.
What to expect
  • STEP 1 – You work with Technical Support to schedule the installation and prepare for downtime
  • STEP 2 – You & your IT team back up methods, raw data, and required files
  • STEP 3 – A Thermo Fisher Field Service Engineer will come onsite to install and configure the new Windows 11 workstation
  • STEP 4 – The Field Service Engineer will transfer calibration files, calibrate the instrument, and verify performance
  • STEP 5 – You will then complete any remaining data migration, validation, and return your instrument to routine use
Microsoft and Windows are trademarks of Microsoft Corporation.