Quantifying Caffeine in Steeped Tea Using UV-Vis
In the realm of food science, precise quantification of key components in consumables is critical. One such component is caffeine, a stimulant found in tea, which requires accurate measurement to ensure product consistency and safety. UV-Visible absorption spectroscopy has proven to be a powerful method for analysis. This article explores the application of UV-Visible spectroscopy in quantifying caffeine in steeped tea, emphasizing the importance of background correction to achieve accurate results.
Understanding UV-Visible Spectroscopy
UV-Visible spectroscopy is a technique that measures the absorbance of light by a sample at different wavelengths. According to Beer’s law, the absorbance is directly proportional to the concentration of the analyte in solution. This method is particularly advantageous in QA/QC environments, where verifying the correct amount of a substance or ensuring contaminants are below acceptable limits is crucial.
However, the complexity of food matrices can pose challenges. Many food samples contain multiple components, leading to overlapping absorption spectra. This additive nature of absorbance can complicate quantification. Separating these components is a solution, but it is often time-consuming and impractical.
UV-Vis spectroscopy instruments offer quick, inexpensive, easy-to-use analysis to verify whether product quality is within specification, especially for beverages like tea, coffee, beer, wine, juices, energy and soft drinks, waters, and other thin liquids, but also can work with sticky and thick liquids, like honey, or oily ones, like palm oil.
Understanding Tea, Steeped or Not
The caffeine content in tea is primarily released during the steeping process. Therefore, tea that has not been steeped will contain negligible amounts of caffeine in the liquid since the caffeine remains in the dry tea leaves.
Here is a comparison for clarity:
  • Steeped Tea: The caffeine content in steeped tea can vary widely depending on factors such as the type of tea, the steeping time, and the water temperature. A search on the internet shows that many tea companies report that on average, an 8-ounce cup of black tea contains about 40-70 milligrams of caffeine, while green tea contains about 20-45 milligrams. Noting the same varying factors, the Mayo Clinic* offers these numbers as a guide: 8 ounces of brewed black tea contains 47 mg of caffeine, while 8 ounces of brewed green has 28.
  • Non-Steeped Tea: Dry tea leaves themselves do contain caffeine, but until they are steeped in hot water, the caffeine is not released into a consumable form. Therefore, non-steeped tea leaves do not contribute measurable caffeine content to a beverage.
If you’re a tea drinker, the Tea Association of the United States advises that for a good cup of black tea, one should steep 3-5 minutes, with other suggested times for green, oolongs, and white teas. If you’re a scientist, however, trying to measure the caffeine in tea, you need some analytical instruments.
Application in Caffeine Quantification
We conducted a test, using a UV-Visible spectrophotometer, for quantifying caffeine in steeped tea.
The process began with the preparation of tea samples, which were steeped for varying durations. As expected, increasing steep times resulted in higher absorbance, indicating more components are extracted from the tea leaves. This was evidenced by the absorption spectra of tea samples steeped for 30 seconds, 2 minutes, 4 minutes, and 10 minutes.
A critical aspect of the method is the background correction, which is essential to remove interference and ensure accurate quantification. Background correction refers to the process of adjusting for any interference or noise that might affect the accuracy of measurements. These extraneous factors could include the type of tea, the water quality, and the quality of the tea itself.
Results and Discussion
When we finished analyzing, the absorption spectra of caffeine standards and tea samples were compared. The standards revealed a clear absorption feature at 272 nm, which was used to quantify caffeine concentration. The absorbance at this wavelength was plotted against caffeine concentration, resulting in a linear relationship.
The tea samples, measured at different steep times, showed corresponding increases in absorbance, aligning with the expected increase in caffeine extraction over time. The absence of significant absorption features beyond 450 nm indicated minimal interference from particulates, affirming the efficacy of the filtration process.
Conclusion
This study demonstrated that accurate and efficient caffeine quantification could be achieved via UV-Visible spectroscopy.
For food scientists, this method offers a non-destructive means to analyze the absorbance spectrum of tea samples, allowing for further analysis if needed, making it a valuable tool in QA/QC environments.
UV-Visible spectroscopy, when combined with advanced background correction methods, provides a powerful approach for the quantification of caffeine in steeped tea.
This technique not only helps ensure product quality and safety but also streamlines the analytical process, making it an indispensable tool for food scientists.
For more information, including study methods and process, instruments used, absorption spectra, equation derivation, derivative analysis comparisons, and caffeine concentration calculation charts, read the application note: Caffeine Quantification in Steeped Tea: Background Correction Using GENESYSTM Smart QC.
Additional Resources
Kyle Mcnally