A Safer, Smarter Alternative for FTIR Spectroscopy: Exploring Thermoelectrically Cooled MCT Detectors
Fourier-transform infrared (FTIR) spectroscopy is a mainstay in materials analysis, catalysis studies, and pharmaceutical research. It’s the ideal technology for detecting weak signals and resolving fine spectral details, particularly in the MIR region (4000–400 cm⁻¹). However, FTIR detectors require cooling to minimize thermal noise and preserve that high sensitivity. This is typically achieved using liquid-nitrogen-cooled detectors, but this poses safety risks, logistical hurdles, and environmental concerns.
Thermoelectrically cooled (TEC) mercury cadmium telluride (MCT) detectors offer an innovative approach that eliminates the need for liquid nitrogen while maintaining high performance. Here, we will explore the risks associated with traditional cooling methods and highlight the key considerations of TEC technology.
The Risks of Liquid Nitrogen in the Lab
Liquid nitrogen, while invaluable in many research settings, comes with several challenges:
  1. Safety Hazards: Liquid nitrogen is cryogenic and can cause severe frostbite or respiratory harm if mishandled. Accidental contact with skin or inhalation of nitrogen gas in confined spaces poses significant risks.
  2. Logistical Challenges: Storing and replenishing liquid nitrogen requires specialized equipment and frequent deliveries, especially in high-consumption laboratories.
  3. Environmental Impact: Although nitrogen is abundant in the atmosphere, the energy-intensive processes used to liquefy and transport it contribute to the lab’s carbon footprint.
  4. Operational Interruptions: Dependency on liquid nitrogen introduces downtime risks, as any disruption in supply can halt analytical workflows.
The TEC MCT Detector: A New Approach to FTIR Analysis
Thermoelectrically cooled MCT detectors provide an innovative solution to many challenges associated with traditional FTIR spectrometry. By replacing liquid nitrogen with a solid-state thermoelectric module, these detectors eliminate the risks and logistical hurdles tied to cryogenic cooling. This shift ensures consistent performance without the operational and safety concerns of handling liquid nitrogen.
Removing liquid nitrogen from the equation not only enhances safety but also significantly improves usability. Researchers no longer need to worry about cryogenic burns or the potential ventilation issues caused by nitrogen gas evaporation. This makes the technology inherently safer and more practical for everyday lab use.
Additionally, TEC MCT detectors are designed to simplify laboratory workflows. By eliminating the dependency on liquid nitrogen deliveries, they are particularly beneficial for labs located in remote areas or those with limited infrastructure. This accessibility allows more institutions to utilize high-performance FTIR analysis without facing logistical barriers.
TEC detectors also represent a meaningful advancement from a sustainability perspective. Reducing reliance on liquid nitrogen decreases the environmental footprint of laboratories by mitigating the energy-intensive processes required for nitrogen liquefaction and transport. This aligns with the growing emphasis on greener and more sustainable research practices.
Applications, Advantages & Limitations
TEC MCT detectors enable safer, more streamlined workflows, addressing common challenges faced by researchers in materials science, chemistry, and environmental monitoring. In the interest of balance, it should be noted that TEC systems may represent a higher upfront cost than traditional detectors, and that traditional liquid nitrogen-cooled systems might still offer performance advantages in select ultra-high-sensitivity applications. However, they are extremely valuable for for applications requiring precision and speed, such as:
  • Catalysis and Reaction Kinetics: Rapid acquisition rates ensure accurate analysis of fast reactions.
  • Gas and Thin Film Analysis: High sensitivity enhances the detection of minute spectral features.
  • Cure Studies and Polymers: Consistent performance allows for precise monitoring of time-sensitive processes.
The Future of Safer and Smarter FTIR Analysis
Thermoelectrically cooled MCT detectors represent a significant advancement in FTIR spectroscopy by addressing traditional cooling methods’ safety and logistical challenges. Their ability to maintain performance while eliminating reliance on liquid nitrogen makes them a compelling option for many research and industrial applications.
As laboratories continue to prioritize safety, sustainability, and efficiency, TEC MCT detectors provide a forward-thinking solution that aligns with these goals. Researchers can choose the technology that best meets their needs by carefully evaluating the advantages and considerations.
Resources
Written by:
Lena Woodis
Product Manager - FTIR, Thermo Fisher Scientific
Lena Woodis, Product Manager at Thermo Fisher Scientific, supports FTIR benchtop instruments and applications. She holds a PhD in physical chemistry has over 15 years of experience in vibrational spectroscopy and chemical instrumentation.
Read more Woodis, Lena