7 Reasons Metal Fabricators Should Analyze Coatings with Handheld XRF Technology
7 Reasons Metal Fabricators Should Analyze Coatings with Handheld XRF TechnologyMetals that are left out in the elements can rust.  And rust or corrosion in general can cause critical failures (think structural metal beams, airplanes, and automobiles.)
The air, rain, snow, salt, even pollutants like sulphur dioxide can adversely affect the metal’s look and durability. To avoid, or at least, reduce the deterioration of the metal, fabricators apply various coatings, depending on the customer’s requirements.  However, applying coatings to metal is an exact science.
How do metal fabricators verify the specification conformance of their coatings? One way is to use handheld XRF technology. XRF is an acronym for X-ray fluorescence spectroscopy. XRF is a non-destructive analytical technique used to determine the elemental composition of materials. Handheld XRF analyzers work by measuring the fluorescent (or secondary) X-rays emitted from a sample when excited by a primary X-ray source. Each of the elements present in a sample produces a set of characteristic fluorescent X-rays, or “unique fingerprints”. These “fingerprints” are distinct for each element, making handheld XRF analysis an excellent tool for quantitative and qualitative measurements.
Handheld XRF analyzers are an extremely efficient technology for rapid compositional analysis of alloys and coating thickness measurements. Since the analyzers are handheld, they can be utilized to perform spot analysis anywhere in the supply chain, help ensure that expensive coating material isn’t wasted, and enhance process monitoring and quality control. 
I’ve outlined below seven reasons to use XRF technology with a coatings mode to analyze metal coatings:
  1. Ensure goods received match purchase orders by measuring metal grade and composition.
  2. Reduce production costs by minimizing production errors.
    • Coatings that are too thin can result in poor corrosion resistance, high warranty costs and/or product failure.
    • Coatings that are too thick give money away by using more product than is necessary.
    • Non-destructive analysis means there is no need to cut or damage high-value product
  3. Improve quality by ensuring consistent coating across a product via multiple measurements and automatic averaging.
  4. Provide faster turnaround with immediate results and no sample preparation (versus statistical sampling followed by lab analysis which is time consuming).
  5. Generate quality reports and certificates through simple report generation tools.
  6. Create audit trails of product from goods incoming inspection to products out the door.
  7. Achieve peace of mind with compliance to international methods ISO 3497 and ASTM B568.
Confirming that coating specifications are met while performing quality control at-line, online or during final product inspection is a necessity.  XRF analysis can be the tool that helps ensure those coatings are correct.
Additional Reading:
Written by:
Mathieu Bauer
Senior Application Scientist, Thermo Fisher Scientific
Mathieu has almost 25 years of experience with various analytical technologies including NIR, FT-IR, ICP-OES, MS, XRF and LIBS. Mathieu is the Senior Application Scientist for the Thermo Scientific Niton Analyzers product line and provides his expertise in handheld XRF and handheld LIBS to our commercial team and end-users. He also supports the development of new markets and applications for both technologies. Mathieu is fluent in French, English and German and is based out of Europe.
Read more Bauer, Mathieu