Renewed Interest in Coal-Fired Power Generation Technologies 
Summary
In June 2026, the U.S. Department of Energy (DOE) announced $350 million investment in four coal modernization and reliability projects aimed at strengthening coal-based generation, enhancing grid reliability, and supporting America’s strategic energy infrastructure. These projects are intended to improve plant efficiency, extend operational lifespans, and reinforce the role of coal-fired power plants in providing reliable baseload power. 
The DOE also announced plans to support 13 U.S. coal-fired power plants and new coal export infrastructure through up to $500 million in Defense Production Act (DPA) Title III funding. 
As interest in coal-fired power generation grows, coal producers and plant operators are evaluating technologies that can improve efficiency, support environmental compliance, and enhance operational decision-making. From coal quality monitoring and blend optimization to emissions measurement and process control, advanced analytical technologies provide valuable insights throughout the power generation process—from the mine to the boiler and ultimately to the electric grid. 
Introduction 
Modern coal-fired power plants rely on much more than boilers, turbines, and generators to produce electricity efficiently. Today’s facilities incorporate advanced technologies for fuel management, process optimization, combustion control, emissions monitoring, and worker safety. 
These analytical and monitoring systems provide real-time information about fuel quality, operating conditions, equipment performance, and environmental compliance. By transforming data into actionable insights, they help operators improve efficiency, reduce operating costs, maximize asset utilization, and maintain reliable power generation. 
Understanding the technologies that support coal-fired power generation provides valuable insight into how modern facilities optimize performance while balancing economic and environmental objectives. 
Coal Quality Analysis and Fuel Management 
The quality of coal delivered to a power plant can vary significantly depending on its source, mining conditions, and processing methods. Variations in ash content, sulfur levels, moisture, and heating value can directly affect combustion efficiency, emissions performance, and operating costs. 
To address these challenges, many facilities employ online coal quality analyzers that continuously measure key fuel characteristics as coal moves through conveyor systems. Unlike traditional laboratory testing, which can require hours or days to generate results, real-time analysis provides immediate feedback that enables operators to adjust blending strategies, fuel handling practices, and combustion parameters before coal enters the boiler. 
Continuous coal quality monitoring helps maintain consistent fuel specifications, improve operational stability, and support more efficient power generation. 
Elemental Analysis and Coal Blend Optimization 
Many coal-fired power plants use elemental analysis technologies to continuously measure the chemical composition of coal, ash, and other process materials. Techniques such as Prompt Gamma Neutron Activation Analysis (PGNAA) and Pulsed Fast Thermal Neutron Activation (PFTNA) provide real-time measurements of elemental concentrations in bulk materials. 
PGNAA and PFTNA are noncontact, non-destructive analytical techniques used in online analysis systems to determine the elemental composition of coal, and help provide coal producers with accurate, reliable data to control coal blends and ensure on-spec batches and more efficient use. 
By measuring sulfur, moisture, ash, calorific value, and other key parameters, these technologies help operators better understand fuel variability and make informed decisions regarding coal blending, combustion optimization, and ash management. 
Advanced blend optimization software further enhances fuel management by continuously monitoring coal quality and creating blend recipes that meet specific operating requirements. These systems can combine coal from multiple sources while targeting parameters such as ash, sulfur, moisture, and heating value. The result is more efficient use of fuel resources, reduced variability, and more consistent boiler performance.  
Metal Analysis for Infrastructure Integrity 
The reliability of a coal-fired power plant depends heavily on the integrity of its piping, pressure vessels, pumps, valves, and other critical infrastructure. Positive Material Identification (PMI) technologies help verify that components are manufactured from the correct alloy and meet design specifications. 
Handheld X-ray Fluorescence (XRF) and Laser-Induced Breakdown Spectroscopy (LIBS) analyzers are commonly used to identify and verify metallic materials. XRF rapidly determines elemental composition using X-ray interactions, while LIBS uses laser-generated plasma to identify elements, including carbon. LIBS is capable of quantifying carbon and is especially suited for positive material identification (PMI) of piping, pressure vessels, valves, pumps, and finished welds, or to grade unknown materials to regain traceability – as well as measuring silicon in steel for sulfidic corrosion or residual elements in HF Alkylation process components. 
These technologies help manufacturers, contractors, and plant operators verify material composition before installation and during maintenance activities, reducing the risk of failures caused by incorrect or out-of-specification materials. 
Air Quality Monitoring for Worker Safety 
Air quality monitoring plays an important role in protecting workers and surrounding communities. Coal mining and power generation operations can generate airborne dust and particulate matter that may affect worker health, workplace conditions, and the surrounding environment. 
Monitoring systems are used to track coal dust exposure, perimeter air quality, fugitive dust emissions, and workplace conditions.  
Personal Dust Monitors help protect miners’ health by tracking the shift-average respirable coal dust exposure as it approaches regulatory limits. By providing mine workers and managers with real-time personal coal dust monitoring, these monitors support timely decision-making and help reduce the risk of long-term health effects associated with dust exposure. Fugitive dust is typically created through activities such as material handling, the physical movement of soil, vehicles traveling over unpaved surfaces, heavy equipment operation, blasting, or wind erosion. Many jurisdictions establish exposure limits or permit conditions for activities likely to generate high levels of particulate matter. Real-time monitoring helps organizations identify potential risks, implement corrective actions, and maintain safe working environments. 
Emissions Monitoring Technologies 
Coal combustion produces emissions that must be monitored and managed to meet environmental regulations and operating requirements. Continuous Emissions Monitoring Systems (CEMS) provide real-time measurement of pollutants and process gases, such as sulfur dioxide (SO₂), nitrogen oxides (NOₓ), carbon monoxide (CO), carbon dioxide (CO₂), oxygen (O₂), particulate matter (PM), and, where required, mercury (Hg). 
These systems enable operators to verify compliance with environmental permits, evaluate the performance of pollution-control equipment, and identify opportunities for process improvements. Accurate emissions monitoring also supports regulatory reporting and helps facilities maintain efficient, environmentally responsible operations. 
Radiation Detection and Monitoring 
Coal and other mined materials naturally contain trace amounts of radioactive elements. In addition, power generation facilities often use X-ray and gamma-ray monitoring technologies for measurement and analytical applications. 
As a result, radiation monitoring is an important component of worker safety and operational control. Radiation detection systems help identify and measure natural and man-made radiation sources throughout the facility. Portable and handheld devices provide real-time monitoring, helping personnel assess potential exposure risks and respond appropriately when elevated radiation levels are detected. 
These technologies contribute to a safer workplace while supporting compliance with radiation safety protocols. 
Conclusion
Modern coal-fired power generation relies on far more than boilers and turbines. Advanced technologies for coal quality analysis, elemental monitoring, blend optimization, material handling, infrastructure inspection, emissions monitoring, air quality assessment, and radiation detection play essential roles in maintaining efficient and reliable plant operations.
By providing real-time information about fuel characteristics, equipment condition, environmental performance, and process efficiency, these technologies help operators make informed decisions across the entire value chain—from the mine to the power plant. As investment in coal modernization continues, analytical and monitoring technologies will remain critical tools for improving efficiency, supporting compliance, and enhancing operational performance.
Frequently Asked Questions
  • Coal-fired power plants use online analyzers, automated sampling systems, and laboratory testing to measure key fuel properties such as ash content, sulfur levels, moisture, and heating value. These technologies provide operators with the information needed to manage fuel quality, optimize combustion, and maintain consistent plant performance.
  • Combustion optimization systems use sensors and process controls to monitor factors such as oxygen levels, temperature, and fuel feed rates in real time. By continuously adjusting operating conditions, these systems help improve heat transfer, reduce fuel consumption, minimize unburned carbon, and increase overall power generation efficiency.
  • Emissions monitoring technologies measure gases released during combustion, including sulfur dioxide (SO₂), nitrogen oxides (NOₓ), carbon monoxide (CO), and carbon dioxide (CO₂). Continuous monitoring helps power plants comply with environmental regulations, evaluate pollution-control equipment performance, and identify opportunities to improve operational efficiency.
  • Real-time data enables operators to make informed decisions quickly. Information from coal analyzers, process instruments, and emissions monitoring systems helps identify changes in fuel quality or operating conditions before they affect plant performance. This proactive approach can improve efficiency, reduce downtime, and support more reliable power generation.
  • Process monitoring technologies continuously track equipment performance, material flow, fuel characteristics, and environmental conditions throughout the plant. By providing early warning of potential issues, these systems help operators prevent unplanned outages, optimize maintenance schedules, and maintain stable plant operations over the long term.
Written by:
Kevin Gordon
Sr. Manager - Applications, Thermo Fisher Scientific
With over 20 years of experience in process control, Kevin Gordon has honed expertise in Prompt Gamma Neutron Activation Analysis (PGNAA) technology.
Read more Gordon, Kevin