How To Correctly Prepare Pellets Before Analyzing Your Mining Sample
proper sample prep mining pellets
Proper sample prep of mining pellets
Gary Thomas, Editor-in-Chief of AZoM, wrote a nice article on how to correctly prepare pellets before analyzing your sample.  He recommends: When samples are prepared for XRF analysis, the size of the particles to be analyzed should be maintained within the saturation depth of the x-rays. A preliminary size reduction is carried out for the sample materials of large feed sizes. Following this, a small part of the sample is subjected to fine grinding. Dry and bulk samples can be introduced into the rotating dividers through vibratory feeders for grinding. Sample splitters are used for heavily flowing materials. Finally, the part sample thus obtained is subjected to pulverization. We agree, especially in the mining industry.  Whatever your mining and exploration requirements are, you need  information that’s accurate, precise, and fast. With the proven analytical performance of portable XRF analyzers in the field at all stages of the exploration, mining, and refining processes, the limiting factor in achieving better correlation with laboratory results is proper sample preparation.  And with proper sample preparation comes proper pellet preparation. Take a look at these 6-minute videos that give step-by-step instructions on how to pelletize rock samples right in the field…using both a  hammer mill field kit and a direct rock sampler kit.
Round gray circle with a human silhouette. Dark gray shadow on the bottom. Isolated on a light background. EPS 10.
Written by:
Jon Culbertson
Former Director of Business Development, Thermo Fisher Scientific
Jon Culbertson spent over 16 years at Thermo Fisher Scientific in various positions, including Director of Business Development (Portable Analytical Instruments Business Unit) where he directed a global team of geologists, X-Ray Fluorescence ("XRF") scientists and business development professionals working with field-portable XRF solutions to the exploration (including oil and gas E&P), mineral processing, precious metals and regulatory (consumer goods, lead paint and environmental) markets.
Read more Culbertson, Jon