How to Visualize Gas Leaks on Pipelines: A Practical Guide Using Optical Gas Imaging
Summary
Gas pipeline leaks present significant challenges for safety, environmental compliance, and operational efficiency. As pipeline networks become longer and more complex, traditional inspection methods are no longer the best fit for quickly and accurately locating leaks. Optical Gas Imaging (OGI) has emerged as a powerful solution, enabling real-time visualization of gas leaks directly on pipelines.
This article provides an overview of how to visualize gas leaks on pipelines using modern OGI techniques, including handheld, fixed, and aerial approaches.
Why Visualizing Pipeline Leaks Matters
Gas pipelines often span large distances and operate under varying environmental conditions. Detecting leaks is not enough—operators must quickly visualize and localize the exact emission source to take corrective action.
Key challenges include:
  • Long-distance infrastructure coverage
  • Limited physical access to elevated or remote sections
  • Environmental interference such as wind and humidity
  • Difficulty identifying exact leak points using traditional tools
OGI technology provides a more effective approach by visualizing gas emissions as colorized plumes, helping operators quickly identify leaks and prioritize remediation efforts.
What Is Gas Leak Visualization?
Gas leak visualization is the process of using infrared imaging to make hydrocarbon gases visible. OGI cameras detect specific wavelengths absorbed by gases such as methane and VOCs, rendering them as colorized plumes on a display.
Instead of relying on numerical readings, operators can:
  • See the leak in real time
  • Identify the exact source location
  • Assess leak size and behavior visually
This approach is especially valuable for pipeline inspections, where speed and accuracy are critical.
Methods to Visualize Gas Leaks on Pipelines
1. Handheld Optical Gas Imaging
Handheld OGI cameras are the most direct way to visualize leaks along pipelines.
How It Works:
  • The operator scans pipeline components such as valves, joints, and connectors
  • The camera displays gas plumes in real time
  • Leaks are immediately localized without additional tools
Advantages:
  • Rapid inspection of accessible pipeline sections
  • High sensitivity to small leaks
  • Safe operation from a distance
Handheld devices are particularly effective for routine inspections and maintenance checks along pipeline routes.
2. Long-Range Visualization for Pipelines
Pipelines often extend across large or hard-to-reach areas. Long-range OGI capabilities allow operators to visualize leaks from significant distances. Detection can happen from tens to hundreds of meters away, reducing the need for direct access to hazardous or elevated pipeline sections.
Key Technologies:
  • Telephoto lenses for extended viewing distances
  • Spectral filtering to reduce atmospheric interference
  • Enhanced sensitivity for detecting emissions under challenging conditions
  • When the long-range lens is used in combination with the heavy alkanes filter, operators can minimize the effects of high humidity and more effectively visualize gas leaks.
3. Drone-Based Pipeline Inspection
Aerial platforms equipped with OGI cameras provide a scalable solution for long-distance pipeline monitoring. Drone-based visualization is especially useful for transmission pipelines, storage tanks, offshore or isolated infrastructure and emergency leak assessments. By capturing real-time imaging from above, operators can quickly identify and localize leaks that would otherwise be difficult to detect.
Benefits:
  • Access to remote or elevated pipeline segments
  • Rapid coverage of large areas
  • Reduced need for manual inspection in difficult terrain
4. Fixed Monitoring Systems for Continuous Visualization
For critical pipeline infrastructure, fixed OGI systems provide continuous monitoring without manual intervention and immediate, automated leak detection. In addition, they can integrate environmental information such as wind direction.
How It Works:
  • Cameras and sensors are installed along key pipeline points
  • The system continuously scans for emissions
  • AI-based analysis identifies abnormal leak patterns
Quantifying Pipeline Leaks After Visualization
Once a leak is visualized, quantification becomes essential for reporting and compliance.
Modern OGI systems can estimate leak rates in real time and automatically record inspection data or generate reports for regulatory requirements. This combination of visualization and quantification transforms pipeline inspections from reactive processes into data-driven operations.
Best Practices for Visualizing Pipeline Leaks
Here are some best practices that can help maximize the effectiveness of OGI technology in visualizing pipeline leaks:
  • Scan systematically along pipeline components and connection points
  • Use long-range capabilities for inaccessible areas
  • Leverage aerial inspections for large-scale coverage
  • Monitor environmental conditions to optimize detection accuracy
  • Combine technologies (handheld, fixed, and drone) for full coverage
A layered approach helps ensure no section of the pipeline is left unmonitored.
Conclusion
Visualizing gas leaks on pipelines is no longer a complex or time-consuming task. Optical gas imaging (OGI) enables operators to detect, localize, and assess leaks in real time, even across large and challenging environments.
By combining handheld devices, long-range imaging, drone inspections, and fixed monitoring systems, organizations can achieve comprehensive pipeline visibility. This not only improves safety and compliance but also reduces operational costs and environmental impact.
As pipeline networks continue to expand, adopting advanced visualization technologies is essential for maintaining efficient, compliant and reliable operations.
You can watch a webinar recording delving more into the subject here: Seeing Gas Leaks in Real Time: How Handheld Optical Gas Imaging Improves LDAR.
  • Gas leaks are visualized using infrared OGI cameras that detect specific wavelengths absorbed by gases, making them visible as plumes on the camera display.
  • Yes, modern OGI systems with telephoto lenses and spectral filters enable long-range leak visualization. Offshore platform leaks have been seen at 10km.
  • Yes, drones provide aerial access to remote or elevated pipeline sections, improving inspection coverage.
  • Wind, humidity, and temperature differences can impact detection, but advanced OGI systems are designed to minimize these effects.
  • Yes, fixed OGI systems allow for 24/7 monitoring and real-time leak detection along critical pipeline infrastructure.
Bob Gallagher
Written by:
Bob Gallagher
Product Line Manager, Thermo Fisher Scientific
Bob Gallagher is a Product Line Manager at Thermo Fisher Scientific, focused on the engineering and product management of industrial hygiene products, including LDAR, site remediation, dust monitoring and other various industrial applications.
Read more Gallagher, Bob