Seeing Gas Leaks in Real Time: How Optical Gas Imaging Transforms LDAR Programs 
Article Summary
Leak Detection and Repair (LDAR) programs are essential for maintaining environmental compliance, operational safety, and cost efficiency across industrial facilities. Traditional gas leak detection methods often fall short in speed, accuracy, and usability. Recent advancements in optical gas imaging (OGI) technology are reshaping how organizations detect, visualize, and quantify gas emissions in real time. This article summarizes key insights from a technical webinar on handheld optical gas imaging and its role in improving LDAR performance, offering a clear, search-optimized overview of how OGI enhances leak detection, compliance, and operational efficiency.
The Evolution of Gas Leak Detection 
Gas leak detection has evolved significantly over the past century. Early methods relied on indirect indicators such as animal behavior or rudimentary techniques like applying soapy water to detect bubbles. These approaches were time-consuming, inconsistent, and often unsafe.  
Later, concentration-based detection tools improved accuracy but still required close proximity to potential leaks and lacked precise localization capabilities.  
Today, optical gas imaging represents a major advancement. By using infrared technology to visualize gas emissions, OGI allows operators to “see” leaks instantly, eliminating guesswork and dramatically improving inspection workflows. 
What Is Optical Gas Imaging (OGI)? 
Optical Gas Imaging (OGI) is a technology that uses infrared cameras to detect hydrocarbon gases and volatile organic compounds (VOCs). These cameras isolate specific wavelengths absorbed by gas molecules, making otherwise invisible emissions visible in real time.  
Key Capabilities of OGI: 
  1. Real-time visualization of gas leaks  
  2. Detection of methane and hundreds of VOCs  
  3. Remote operation for improved safety  
  4. Rapid inspection of large facilities 
Unlike traditional methods, OGI enables operators to immediately pinpoint the exact source of a leak without relying solely on concentration readings. 
Why OGI Improves LDAR Programs 
1. Faster Leak Detection and Localization 
OGI cameras allow inspectors to scan equipment quickly and identify leaks instantly. This significantly reduces inspection time and improves maintenance response.  
2. Enhanced Safety Through Remote Operation 
Because OGI systems operate at a distance, technicians avoid direct exposure to hazardous gases or difficult-to-access equipment, improving overall workplace safety.  
3. High Sensitivity for Early Detection 
Advanced OGI systems are a nice complement to the Method 21 analyzers, such as a toxic vapor analyzer, which together can detect extremely small leaks, enabling early intervention before issues escalate into safety or environmental risks.  
4. User-Friendly Operation 
Modern OGI devices are designed for ease of use, allowing operators to perform inspections with minimal training while maintaining high accuracy. 
Advanced Features of Modern OGI Cameras 
Multi-Gas Detection and Spectral Filtering
Modern OGI systems can detect a wide range of gases by switching spectral filters in just a few minutes. This capability enhances flexibility across different industrial applications and environments.  Additionally, optimized filters improve performance in challenging conditions such as high humidity or long-distance inspections.
Built-In Quantification for Reporting
Quantification tools allow users to estimate leak rates directly from captured data. This is critical for regulatory compliance and emissions reporting, supporting data-driven decision-making. 
Self-Calibration and Reliability
Newer systems incorporate self-calibration features that verify detection accuracy each time the device is used. This helps ensure consistent performance and reduces the need for manual calibration processes.  
Extended Battery Life and Operational Efficiency
Long battery life enables continuous inspections over full work shifts, reducing downtime and improving productivity. 
Expanding LDAR with Integrated Monitoring Solutions 
While handheld OGI cameras are powerful tools, large facilities often require broader monitoring strategies. 
Fixed Monitoring Systems 
Fixed OGI systems provide continuous, site-wide monitoring by integrating multiple detection technologies, including cameras, sensors, and environmental data inputs.  
These systems: 
  • Detect abnormal emission patterns  
  • Learn site-specific behavior using AI-driven analytics  
  • Provide real-time alerts and leak localization  
Drone-Based Inspections 
Aerial platforms equipped with OGI cameras extend coverage to hard-to-reach areas such as pipelines, storage tanks, elevated structures, and remote infrastructure.  
Integrated Detection Ecosystem 
Combining handheld devices, fixed systems, and drone platforms creates a comprehensive monitoring approach that maximizes coverage and minimizes undetected emissions.
The Role of Regulation in OGI Adoption
Environmental regulations are a primary driver of OGI adoption worldwide. Compliance frameworks require accurate detection, quantification, and reporting of emissions. 
OGI technology supports compliance by meeting regulated detection sensitivity thresholds, helping to enable accurate emissions quantification, and providing audit-ready inspection data.
As regulations continue to evolve, advanced OGI systems are becoming essential tools for meeting both current and future requirements.
Operational and Cost Benefits of OGI 
Beyond compliance, OGI delivers measurable operational advantages: 
  • Reduced inspection time and labor costs  
  • Lower maintenance expenses through early detection  
  • Minimized product loss from undetected leaks  
  • Improved environmental performance  
By preventing leaks and optimizing inspection workflows, OGI helps contribute to long-term cost savings and operational efficiency. 
Conclusion
Optical Gas Imaging is transforming how industries approach leak detection and environmental monitoring. By enabling real-time visualization, precise localization, and reliable quantification of gas emissions, OGI helps to significantly enhance LDAR programs. 
As regulatory pressures increase and operational efficiency becomes more critical, integrating handheld, fixed, and aerial OGI solutions offers a scalable and future-ready approach to emissions management. Organizations that adopt these technologies position themselves to improve safety, reduce costs, and achieve higher standards of environmental compliance. 
You can watch a webinar recording delving more into the subject here: Seeing Gas Leaks in Real Time: How Handheld Optical Gas Imaging Improves LDAR. 
  • OGI allows users to visually detect gas leaks in real time, making it faster to catch medium to large leaks in between quarterly monitoring periods.   
  • Yes, modern OGI systems are highly sensitive and are a nice complement to the Method 21 analyzers, such as the Thermo Scientific™ TVA2020, which, when used together, can detect extremely small leaks, enabling early intervention before issues escalate into safety or environmental risks.  
  • Yes, especially when combined with fixed monitoring systems and drone-based inspections for full-site coverage. 
  • OGI provides accurate detection and quantification data, helping organizations meet emissions reporting and environmental standards. 
  • Advanced systems include self-calibration features, reducing the need for frequent manual calibration. 
Bob Gallagher
Written by:
Bob Gallagher
Product Line Manager, Thermo Fisher Scientific
Bob Gallagher is a Product Line Manager at Thermo Fisher Scientific, focused on the engineering and product management of industrial hygiene products, including LDAR, site remediation, dust monitoring and other various industrial applications.
Read more Gallagher, Bob