Infographic: Hearing a “Tin Cry”
Tin InfographicThis metal is never found as the free element; the most important ore is cassiterite (tin oxide, SnO2).  It has a highly crystalline structure and causes a “tin cry” to be heard when a bar is bent.  So it’s no surprise that this mined metal, which traditionally has been used to celebrate 10th anniversaries, is Tin. Read more about this malleable metal in this infographic: 9 Fast Facts About Tin.