  • Contact
Accelerating ScienceAdvancing Mining / Exploration / Technical Poster - Application of Portable XRF Analyzer in Geochemical Exploration
Technical Poster – Application of Portable XRF Analyzer in Geochemical Exploration
Written by Ali Somarin | Published: 09.03.2013
Technical Poster: XRF Analyzer in Geochemical Exploration
Technical Poster: XRF Analyzer in Geochemical Exploration
This technical poster was presented at St. John’s 2012 GAC-MAC joint annual meeting . [The Geological Association of Canada (GAC®) and the Mineralogical Association of Canada (MAC)]
Application of the Thermo Scientific Niton portable XRF analyzer in geochemical exploration: An example from the Francisco I. Madero Zn–Pb–Cu–(Ag) deposit, Zacatecas, Mexico
Francisco I. Madero is a Zn–Cu–Pb–(Ag) deposit owned by Peñoles and operated as an underground mine in Zacatecas, central Mexico. Historical production up to 2011 is 23.1 Mt of ore containing 12.8 MOz Ag, 92.09 kt of Pb, and 623.9 kt of Zn.
The deposit consists of several mineralized zones located around a dome-type structure (laccolith) probably generated by forceful emplacement of an intrusive body at depth which shows a wide magnetic and gravimetric anomaly; however, the only intrusive rocks found in the mineralized area are a few Tertiary, post-Laramide dikes. The mineralized area is ~10 km2 with several ore bodies located in the same stratigraphic unit from 30 to 690 m in depth. The stratabound ore bodies are hosted by the Mesozoic back-arc marine sedimentary rocks, suggesting a syngenetic submarine exhalative genesis as sedimentary exhalative (SEDEX) or volcanogenic (VMS). However, calc-silicate mineralogy and replacement textures within calcareous units, and the absence of exhalites or identifiable feeder zones favors distal skarn model.
There are two types of sulfide ore assemblages in the ore body:
  1. Pb-Zn sulfides as NW trending 6-65 m thick masses composed of bands and laminations of sphalerite and galena cut by quartz, clay-pyrite and chlorite-epidote veins at the base of the ore body in an area of 6 km2, and
  2. Cu-Ag sulfide assemblage consists of chalcopyrite, pyrite, cubanite, enargite, and tetrahedrite as laminations and bands in 3-40 m thick ore masses cut by quartz-pyrite-chalcopyrite veinlets.
To compare assay results from different analytical methods and investigate application and efficiency of portable x-ray fluorescence (XRF), three types of analyses were carried out on drill core samples. These methods include in-house Madero atomic absorption (AA), ALS inductively coupled plasma emission spectroscopy (ICP-ES), and Thermo Scientific™ Niton™ portable XRF (Niton XL3t and Niton FXL on pulp samples).
The study shows high correlation between data from portable XRF and lab methods. Cu correlation between XRF-Niton XL3t and ICP method is 0.92 whereas this correlation between XRF-Niton FXL and AA method is 0.96. Similar correlation for Zn assayed by the same instruments yield 0.983 and 0.982 for the core samples of this ore deposit. This correlation drops to 0.87 for Ag. Lead shows the highest correlation (0.99) among the elements that were analyzed by portable XRF and AA or ICP methods.
Systematic analyses of core samples by these three methods and comparative studies indicate that geochemical anomalies for metals of interest in this deposit (Zn-Pb-Cu-Ag) can be identified readily in real time using portable XRF in the field.
View the Technical Poster showing the Application of Portable XRF Analyzer in Geochemical Exploration
Share this article
FacebookLinkedinTwitterMail
Cluster of colorful Fourth of July fireworks
Remarkable Minerals Make Spectacular Fireworks
According to the United States Geological Survey (USGS), min... Marlene Gasdia-Cochraneby / 07.01.2025
Read More
Valves on the pipe
How to Use In-line Raman for Real-Time Natural Gas Compositional Analysis 
Why Raman spectroscopy for natural gas analysis?  Raman spe... by Mayte McHugh / 06.24.2025
Read More
Computer as book eleaning concept - laptop with alphabet letters idea
Did You Miss Our Most Popular Articles? Here are our Top 5 Mining Articles This Year.
Here are the ten most read articles on this mining blog duri... by Marlene Gasdia-Cochrane / 12.18.2024
Read More
Shield design bundle element vector idea with creative modern style
What are the Largest Diamonds and How Are they Found?
In previous articles, we wrote about the January 1905 discov... Marlene Gasdia-Cochraneby / 08.23.2024
Read More
cement ebook
eBook: A Practical Guide to Improving Cement Manufacturing
Download Guide
Mining ebook
eBook: A Practical Guide to Improving Mining and Mineral Operations
Download Guide
Leave a Reply Cancel reply
Your email address will not be published. Required fields are marked *
Did You Miss Us At the Mining Shows?
Anions and Metals Analysis in Hydraulic Fracturing Waters from Marcellus Shale Drilling Operations
Privacy StatementTerms & ConditionsLocationsSitemap
© 2025 Thermo Fisher Scientific. All Rights Reserved.
Talk to us
Notifications
Get news and research reviews on the topic of your choice, right in your inbox.
Subscribe Now
  • This field is for validation purposes and should be left unchanged.
×
  • Tweet
  • Facebook
  • Tweet
  • Facebook