Watson LIMS Software 
Thermo Scientific Watson LIMS Software – built for bioanalysis, trusted by industry leaders. 
For more than two decades, Watson LIMS Software has been the cornerstone of precision, accuracy, and reliability in bioanalytical labs worldwide. It is designed for enhanced traceability, compliance, and control from the first method development to the final study completion. 
With the bioanalysis software, you can summarize assay performance results and study sample results, including the assembly of datasets into SEND and SDTM formats. In addition, Watson LIMS Software lets you emulate study and amendment protocols, providing assurance that what occurs in the system is in exact alignment with the study protocol. 
Watson LIMS Software video overview
Watson LIMS Software capabilities
Manage and build study designs
Upload sample manifests using templates with the aid of Import Study Protocol to reduce metadata transcription errors.
Maintain sample chain of custody
Record all sample shipments and monitor each sample movement, define storage locations down to plate position, and track freeze/thaw cycle and sample stability. Easily provide chain of custody records to auditors. 
Connect lab instruments and software
Reduce the risk of manual transcription errors by automating data capture and management processes. Watson LIMS Software integrates easily with commonly used third party instrumentation through established interfaces (e.g., Sciex Analyst software) and Thermo Fisher CDS software (e.g., Thermo Scientific Chromeleon CDS).
Streamline method validation and sample analysis
Conduct concentration evaluations for pharmacokinetics (PK)/ anti-drug antibodies (ADA) assessments, establish acceptance standards, and highlight results exceeding the range. Mitigate the risk of over- or under-reporting final concentration data through Internal Standard Review.
Execute efficient reporting and QA review
Generate a full study report including method validation experiments, assay performance, and study data. Report your data in CDISC, SEND and SDTM formats to help streamline submissions. 
Ensure traceability throughout a study
Get complete study records and reports from study initiation through study closeout, including sample chain of custody, assay verification, and reconstruction events using the audit trail.  
Get built-in security and audit trail
Benefit from maximum flexibility and configurability while preserving data integrity. System and study access are granted to authorized role-based users only, in accordance with Good Laboratory Practices (GLP). 
Simplify compliance 
Create labels according to study requirements, and display and barcode storage facilities to ease identification and traceability, enabling audit compliance with 21 CFR Part 11, FDA Bioanalytical Method Validation, and EMA guidance. 
Streamline data processing and reporting
Easily generate a variety of graphs, tables, and statistical calculations from study data. Easily sort, find, filter, and reorder grid contents, for a quick way to review a specific sample’s data. 
Minimize quality review time
Get real-time snapshots of data quality and easily transfer study data between sponsor and CRO due to pre-defined assay performance and summary tables.  
Advanced Red Thread analytics
With version 7.7.2, Watson evolves—delivering stronger security, faster validation, and a foundation for the Next-Gen LIMS era. Watson LIMS Software provides enhanced functionality when used with optional Red Thread® modules—Red Thread Fit™ for regression curve fitting, Red Thread Verge™ for ADA cut-point analysis, and Red Thread Eval™ (coming soon) for study output validation—delivering an advanced, audit-ready analytics experience.
From global pharma to agile biotech and CROs, Watson LIMS Software empowers scientists to work with confidence, accelerate decisions, and drive productivity—today and into the future.
Our general purpose product lines are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks.
For General Lab Use Only - Not For Diagnostic Procedures.