Search
After IHC staining of the target antigen, a second chemical stain is often applied to provide contrast that helps the primary staining product stand out. Many of these chemical counterstains show specificity for discrete cellular compartments or antigens, while others will stain the whole cell. The chemical and fluorescent dyes available as counterstains for IHC provide enough variety such that researchers can easily achieve their imaging goals.
The two images that follow provide examples of chromogenic and fluorescence IHC staining, respectively.
CD138 antibody in IHC (FFPE). Chromogenic IHC analysis of Syndecan-1/CD138 (Cat. No. 36-2900) showing staining in the cytoplasm and membrane of FFPE human small intestinal tissue (right) compared to a negative control without primary antibody (left). To expose target proteins, antigen retrieval was performed using 10 mM sodium citrate (pH 6.0), which was heated by microwave oven for 8 to 15 minutes. Following antigen retrieval, endogenous peroxidase activity was blocked in 3% H2O2-methanol for 15 minutes at room temperature. The slide was washed with ddH2O and PBS, and then probed with a Syndecan-1/CD138 polyclonal antibody (Cat. No. 36-2900), diluted in 3% BSA-PBS at a dilution of 1:100 overnight at 4°C in a humidified chamber. Tissues were washed extensively in PBST and detection was performed using an HRP-conjugated secondary antibody followed by colorimetric detection using a DAB kit. Tissues were counterstained with hematoxylin and dehydrated with ethanol and xylene to prepare for mounting.
Fluorescence IHC detection of cytokeratin 18 in colon carcinoma tissue. Human colon carcinoma sections were stained for cytokeratin 18 using an anti–cytokeratin 18 primary antibody (Cat. No. MA5-12104) and Invitrogen DyLight 594–conjugated goat anti-rabbit secondary antibody (Cat. No. 35560, red fluorescence). Thermo Scientific Pierce Hoechst fluorescent stain (Cat. No. 62249) was also used to counterstain the cell nuclei (blue fluorescence).
Hematoxylin, which is usually used together with eosin (H&E stain), is one of the most commonly used dyes in diagnostic histology. It is also used alone as a nuclear counterstain in IHC. Oxidized hematoxylin is combined with aluminum ions to form an active metal–dye complex that stains the nuclei of mammalian cells blue by binding to lysine residues on nuclear histones. This is in contrast to other nuclear stains that label the nucleic acids. The blue/purple color of the nucleus provides good contrast to green, brown, and red stains.
Eosin, which is a brominated fluorescein derivative, is a general cytosolic stain that acts as a non-nuclear counterpart to hematoxylin. Due to its similarity to fluorescein, eosin staining can also be measured via fluorescence microscopy, with max emission of 546 nm. In brightfield, eosin stains bulk tissue a pink to red color, giving good contrast with blue, purple, brown, and green IHC stains.
Nuclear fast red, also called Kernechtrot dye, is another nuclear stain. The main difference between this dye and hematoxylin is that Nuclear fast red stains nucleic acids red in only about 5 minutes, while optimum staining with hematoxylin can take up to 1 hour. Nuclear fast red provides good contrast against blue, purple, brown, and green stains.
Methyl green is another nucleic acid dye that rapidly stains the nuclei green, typical incubation times are 5 minutes. The green coloration provides excellent contrast against blue, purple, brown, and red stains.
Proper selection of nuclear and tissue counter stains is important to maintain optimal color contrast with your desired IHC stains.
Learn more: IHC Troubleshooting Guide
DAPI (4', 6-diamidino-2-phenylindole) and Hoechst 33342 are common nuclear dyes used for fluorescence IHC, as they intercalate into the DNA to give nuclei a strong blue color emission under UV excitation.
Propidium iodide is another nucleic acid dye that is frequently used to stain the nucleus, although with a red emission.
Phalloidin is a fungal toxin that binds to filamentous, but not globular (free) actin. This peptide derivative is commonly conjugated to fluorophores and used in fluorescence microscopic applications to label the actin cytoskeleton in cells. The color used is dependent upon the fluorophore that is conjugated, and a variety of fluorophore-conjugated phalloidin products are commercially available.
Learn more: Cell Staining Tool
Explore: Molecular Probes Cell Imaging Reagent Guide and Protocols App
Type | Dye | Target | Color |
Chemical stain | Mayer’s Hematoxylin | Nuclei | Blue to violet |
Chemical stain | Nuclear fast red (Kernechtrot) | Nucleic acids | Red |
Chemical stain | Methyl green | Nucleic acids | Green |
Chemical stain | Eosin | Cytoplasm | Pink to red |
Fluorescent | Hoechst stain | Nucleic acids | Blue |
Fluorescent | 4', 6-diamidino-2-phenylindole (DAPI) | Nucleic acids | Blue |
Fluorescent | Propidium iodide | Nucleic acids | Red |
Fluorescent | Fluorophore-tagged phalloidin | Filamentous actin | Fluorophore-specific |