Search
For ease of selection, our products are organized into three general categories: Standard manual cryo-tubes, automation-friendly tubes, and automation-compatible open top storage tubes for non-cryo storage applications.
| Standard manual cryotubes |
Automation-friendly cryotubes |
Automation-friendly, open top storage tubes to use with Septa Caps |
|
|
![]() |
![]() |
|
| Product lines |
|||
|---|---|---|---|
| Nalgene, Nunc | Matrix & Nunc Universal ScrewTop Tubes | Matrix Open Top | |
Description |
|||
| Ideal for manual sample processing and storage in standard cryobox format. |
Designed for scalable and automation-friendly processes, and compatible with multiple barcoding technologies for reliable sample ID and tracking. |
Automation - friendly open top storage tubes for use in ambient to cold storage range. Allows for repeated piercing while maintaining seal integrity. |
|
Typical sample storage applications |
|||
Cells Cell culture by-product Biological fluids and derivatives Tissues Genomic materials Proteins |
Cells Cell culture by-product Biological fluids and derivatives Tissues Genomic materials Proteins Small molecules |
Small molecules Genomic materials |
|
| Barcoding |
|||
| 1D (side) + Human readable code (side) |
2D (bottom) 1D + Human readable (side)* 2D Human readable (bottom)* |
2D (bottom) 1D + Human readable (side)* Alpha Numeric |
|
| Sizes |
|||
| 1mL to 15 mL |
0.2 mL to 12 mL |
0.5mL, 0.75mL, 1.4mL |
|
| Storage format |
|||
| Cryobox |
24-, 48-, 96 way ANSI/SLAS rack Cryobox |
96 way ANSI/SLAS rack |
|
| Temperature range |
|||
| Frozen samples to Vapor Phase Liquid Nitrogen** |
Frozen samples to Vapor Phase Liquid Nitrogen* | Ambient to -80°C* |
|
*Temperature ranges are based on the material of the DuraSeal and SepraSeal caps. Minimum temperature of the material of DuraSeal caps is approved to -80°C, SepraSeal caps is approved to -60°C
** Tubes can also be used in liquid nitrogen when protected by CryoFlex