Search
Nalgene plastic funnels are a safer alternative to glass. They won’t break, chip or shatter amid the bustle and bumps of everyday lab work, and won't dent or corrode like metal funnels. Enjoy their light weight and ease of handling.
Trust Nalgene funnels to be made from the highest quality laboratory-grade polypropylene copolymer (PPCO), high density polyethylene (HDPE), low density polyethylene (LDPE), and polypropylene (PP) materials for dependably low extractables and compatibility with a broad range of lab materials.
With so many sizes and styles, there are Nalgene plastic funnels to serve a wide variety of lab applications.
| Nalgene PPCO Long Stem Analytical Funnels | Nalgene PPCO Powder Funnels | Nalgene PPCO Utility Funnels | Nalgene HDPE Large Funnels | Nalgene LDPE Heavy-Duty Funnels | Nalgene PPCO Büchner Funnels |
| Classic long stem design | Classic wide bore stem design | Wide tapered funnel to stem transition | Large size, classic design | Super rugged for rough and tough work conditions | Classic Büchner design in shatterproof plastic |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| Use with standard filter papers for analytical techniques | Use in transferring powder, crystalline chemicals for reagent prep | Use for transferring liquid, pastes, small solids, granular/powder, or viscous samples, reagents | Use for transferring large volumes of liquids | Toughest funnels available for heavy-duty use | Replaces ceramic Büchner funnels for solid sample prep, treatment. Two-piece design, easy to clean |
| External ribbing prevents air lock | External ribbing prevents air lock | Not ribbed | External/internal ribbing prevents airlock | Ribbed | Not ribbed |
| Top diameter 34mm - 158mm | Top diameter 65mm - 147mm | Top diameter 65mm - 155mm | Top diameter 201mm - 258mm | Top diameter 101mm - 248mm | Top diameter 59mm - 172mm |
| Fits standard size filter papers | Fits standard size filter papers | ||||
| PPCO | PPCO | PPCO or HDPE | HDPE | LDPE | PP |
| Milky white translucent | Milky white translucent | Milky white translucent | Milky white | Milky white | Opaque white |
| Temp range -40oC to 121oC | Temp range -40oC to 121oC | Temp range PPCO: -40oC to 121oC HDPE: -100oC to 120oC | Temp range -100oC to 120oC | Temp range -76°C to 70°C | Temp range 0oC to 135oC |
| Autoclavable | Autoclavable | PPCO autoclavable HDPE is not | Do not autoclave | Do not autoclave | Autoclavable |
| Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CFR 177.1520 for food use1 | Complies with Part 21 CF 177.1520 for food use 1 | Complies with Part 21 CFR 177.1520 for food use |
| BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free | BPA-Free |
| Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free | Phthalate-free |
| Nalgene PPCO Long Stem Analytical Funnels | Nalgene PPCO Powder Funnels | Nalgene PPCO Utility Funnels | Nalgene HDPE Large Funnels | Nalgene LDPE Heavy-Duty Funnels | Nalgene PPCO Büchner Funnels | |
| Acids, dilute or weak | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Acids^ strong, concentrated | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Alcohols, aliphatic | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Aldehydes | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Bases/alkali | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Esters | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Hydrocarbons, aliphatic | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Hydrocarbons, aromatic | — | — | — | — | — | — |
| Hydrocarbons, halogenated | — | — | — | — | — | — |
| Ketones, aromatic | — | — | — | — | — | — |
| Oxidizing agents, strong | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| ✓ | ✓ | ✓ | ✓ | ✓ |
| ✓ | ✓ | ✓ | ✓ | ✓ |
| — | — | — | — | — |
1. HDPE and LDPE are acceptable for: Nonacid, aqueous products; may contain salt, sugar or both (pH above 5.0). Dairy products and modifications; oil-in-water emulsions, high or low fat. Moist bakery products with surface containing no free fat or oil. Dry solids with the surfaces containing no free fat or oil (no end-test required) and under all conditions as described in Table 2 of FDA Regulation 177.1520 except condition A - high temperature sterilization (e.g. over 100° C / 212° F)
+Ratings are for short term intermittent exposure. Wash thoroughly between uses.
^Except oxidizing acids; for oxidizing acids, see “Oxidizing agents, strong”
Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.