Technical Resources - Media Formulations
Mammary Epithelial Growth Supplement (MEGS)
A sterile, concentrated (100X) solution intended for use as one component in a complete culture environment for the growth of human mammary epithelial cells. Below is the composition list. The concentrations are proprietary.
ComponentsMolecular WeightConcentration (mg/L)mM
Proteins
EGF (human recombinant)confidentialn/a
Insulinconfidentialn/a
Other Components
Bovine Pituitary Extractconfidentialn/a
Hydrocortisoneconfidentialn/a
Verweis