Mammary Epithelial Growth Supplement (MEGS)
Mammary Epithelial Growth Supplement (MEGS)
Mammary Epithelial Growth Supplement (MEGS)
Gibco™

Mammary Epithelial Growth Supplement (MEGS)

A sterile, concentrated (100X) solution intended for use as one component in a complete culture environment for the growth ofRead more
Have Questions?
Catalog NumberQuantity
S01555 mL
Catalog number S0155
Price (USD)
106.00
Each
Quantity:
5 mL
Price (USD)
106.00
Each
A sterile, concentrated (100X) solution intended for use as one component in a complete culture environment for the growth of human mammary epithelial cells. Each 5 ml bottle of MEGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of mammary epithelial cells and is the correct amount of supplement for a 500 ml bottle of Medium 171.

When a 500 ml bottle of medium is supplemented with MEGS, the final concentrations of the components in the supplemented medium are:

• Bovine pituitary extract (BPE) (0.4% v/v)
• Recombinant human insulin-like growth factor-I (0.01 μg/mL)
• Hydrocortisone (0.5 μg/ml)
• Human epidermal growth factor (3 ng/ml)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeEpithelial Cells (Mammary)
Concentration100 X
Product LineCascade Biologics
Quantity5 mL
Shipping ConditionDry Ice
SpeciesHuman
Product TypeGrowth Supplement
SterilitySterile
Unit SizeEach
Contents & Storage
Store at -20°C in a non-frost-free freezer.

Frequently asked questions (FAQs)

Is Mammary Epithelial Growth Supplement (MEGS) suitable for MCF10A culture?

Yes, Mammary Epithelial Growth Supplement (MEGS) is suitable for MCF10A culture. You can also include 5% horse serum if desired.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.