Technical Resources - Media Formulations
24020 - HBSS, calcium, magnesium
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Calcium Chloride (CaCl2) (anhyd.)111.0140.01.2612612
Magnesium Chloride (MgCl2-6H2O)203.0100.00.49261084
Magnesium Sulfate (MgSO4-7H2O)246.0100.00.40650406
Potassium Chloride (KCl)75.0400.05.3333335
Potassium Phosphate monobasic (KH2PO4)136.060.00.44117647
Sodium Bicarbonate (NaHCO3)84.0350.04.1666665
Sodium Chloride (NaCl)58.08000.0137.93103
Sodium Phosphate dibasic (Na2HPO4) anhydrous142.048.00.33802816
Other Components
D-Glucose (Dextrose)180.01000.05.5555553
Phenol Red376.410.00.026567481
Verweis
1. Hanks, J.H. and Wallace, R.E. (1949) Relation of Oxygen and Temperature in the Preservation of Tissues by Refrigeration. Proc. Soc. Exp. Biol. Med., 71:196.
Related Products:
14025 - HBSS, calcium, magnesium, no phenol red
Cat No: 14025050 , 14025076 , 14025092 , 14025100 , 14025126 , 14025134
14065 - HBSS, 10x, calcium, magnesium, no phenol red
Cat No: 14065049 , 14065056 , 14065072