HBSS, calcium, magnesium
Gibco™

HBSS, calcium, magnesium

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation,Read more
Have Questions?
Catalog NumberQuantity
24020117500 mL
Catalog number 24020117
Price (USD)
31.65
Online exclusive
34.12
Save 2.47
Each
Quantity:
500 mL
Customize this product
Price (USD)
31.65
Online exclusive
34.12
Save 2.47
Each

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.


This HBSS is modified as follows:
With
• Calcium
• Magnesium
• Glucose
• Phenol Red


The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialHanks' Balanced Salt Solution (HBSS)
IncludesNo Sodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 - 305 mOsm/kg
Recommended StorageStorage conditions: 15-30° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
SterilitySterile-filtered
Concentration1 X
For Use With (Application)Variety of Cell Culture Applications; Such as Washing Cells Before Dissociation; Transporting Cells or Tissue Samples; Diluting Cells for Counting; and Preparing Reagents
Physical FormLiquid
Product LineGibco
Quantity500 mL
Solution TypeHank's Balanced Salt
pH6.7 - 7.8
Unit SizeEach

Frequently asked questions (FAQs)

Can I store Hank's Balanced Salt Solution (HBSS) in the refrigerator?

The recommended storage condition for our HBSS products is at 15-30 degrees C. Generally, storage at 4 degrees C will not impact these products. The only concern would be for the 10x HBSS solutions with calcium and magnesium as these salts might precipitate when stored outside the recommended temperature ranges. As long as you don't see any precipitation upon refrigeration, the product will be fine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Cell docking and on-chip monitoring of cellular reactions with a controlled concentration gradient on a microfluidic device.
Authors: Yang Mengsu; Li Cheuk-Wing; Yang Jun;
Journal:Anal Chem
PubMed ID:12199565
We have developed a microfluidic device for on-chip monitoring of cellular reactions. The device consists of two primary analytical functions: control of cell transport and immobilization, and dilution of an analyte solution to generate a concentration gradient. In this device, a dam structure in parallel to the fluid flow was ... More