Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The Human Interferon alpha Antibody (Hu IFN alpha) ELISA quantitates Hu IFN alpha in human serum, plasma or urine samples. The assay will exclusively recognize both natural and recombinant Hu IFN alpha.
Principle of the method
The Human IFN alpha solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Target information
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
IFN alpha (Interferon alpha) is a Type I interferon belonging to a family of structurally related, species-specific proteins exclusive to vertebrates. The interaction of IFN-alpha with its receptor component result in the activation of a number of signaling pathways which are mainly involved in innate immune response against viral infection. In humans, the gene for IFN alpha is located on on chromosome 9. The type I interferons consist of 14 different alpha isoforms (subtypes with slightly different specificities), and single beta, omega, epsilon and kappa isoforms. Homologous molecules are found in many species, including rats and mice (and most mammals), and have been identified in birds, reptiles and fish species. IFN alpha is produced primarily by plasmacytoid dendritic cells, is a potent component of the anti-viral innate immune response and modulates adaptive immunity. Diseases associated with the dysfunction of IFN alpha include Hepatitis C and Hepatitis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : IFL, IFN, IFN-ALPHA, IFN-alpha-2, IFN-alphaA, IFN-alphaD, IFNA, IFNA13, IFNA2, IFNA2A, IFNA2B, IFNA2C, IFNA@, leIF A, leIF D
Gene ID : (Human) 3439, (Human) 3440
Gene symbol : IFNA1, IFNA2
Protein Aliases : alpha-2a interferon, cytokine, IFN-alpha-1/13, IFN-alpha-2, IFNA2, IFNA2b, interferon alpha 1b, interferon alpha 2a, interferon alpha 2b, interferon alpha A, interferon alpha-1/13, Interferon alpha-2, Interferon alpha-A, interferon alpha-D, interferon-alpha1, LeIF A, LEIF A, MGC138207, MGC138505, MGC138507, IFNα, Interferon alpha 13 (IFNA13), Interferon alpha leukocyte (IFL), Interferon alpha1 (IFNA1), Interferon alpha-1/13 (IFN-alpha-1/13), Interferon alpha-D (IFN alphaD), IFNA2B, IFNA2C, IFNAalphaIFN-alpha-1/13, IFNalpha, IFN-ALPHA-2, IFN-ALPHAA, ifa, IFN alpha 1/13, IFN alpha 1b, IFN α, IFNA, IFNA2A, unnamed protein product
UniProt ID (Human) P01563
If an Invitrogen™ immunoassay doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support