Invitrogen
Thermo Fisher technical specialists can help you select the right assay, create a custom assay, and more. Contact us
Analytical sensitivity
45 pg/mL
Assay range
49.15-12,000 pg/mL
Sample type/volume
Plasma
33 µL
Serum
33 µL
Supernatant
100 µL
Hands-on time
1 hr 20 min
Time-to-result
4 hr 45 min
Homogenous (no wash)
No
Interassay CV
<12%
Intraassay CV
<10%
Instrument
Colorimetric Microplate Reader
Product size
96 Tests
Contents
Pre-coated 96 well plate
Standard
Assay Diluent concentrate
Biotinylated Detection Antibody
SAV-HRP
Wash Buffer
Chromogen
Stop Solution
Adhesive Plate Covers
Shipping condition
Wet or Dry Ice
Storage
2-8°C
Protein name
EpCAM (CD326)
Protein aliases
Adenocarcinoma-associated antigen, CD326, Cell surface glycoprotein Trop-1, EGP, EGP314, Ep-CAM, Epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, human epithelial glycoprotein-2, KS 1/4 antigen, KSA, Major gastrointestinal tumor-associated protein GA733-2, membrane component, chromosome 4, surface marker (35kD glycoprotein), Tumor-associated calcium signal transducer 1
Protein family
Other Proteins
Species (tested)
Human
Assay kit format
Sandwich ELISA Kit
Detector antibody conjugate
Biotin
Label or dye
HRP
Gene aliases
DIAR5, EGP-2, EGP314, EGP40, EPCAM, ESA, GA733-2, HNPCC8, KS1/4, KSA, M1S2, M4S1, MIC18, MK-1, TACSTD1, TROP1
Gene ID
Gene symbol
EPCAM
UniProt ID
The Human TROP1/EpCAM ELISA quantitates Hu TROP1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TROP1.
Principle of the method
The Human TROP1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Ep-CAM (epithelial adhesion molecule, epithelial specific antigen, ESA) is a transmembrane glycoprotein expressed in the epithelium with a molecular weight of approximately 40 kDa, which functions as an epithelial cell adhesion molecule. Ep-CAM functions as a homotypic calcium-independent cell adhesion molecule, and has a direct impact on cell cycle, proliferation and metabolism of epithelial cells and fibroblasts due to its ability to rapidly induce the proto-oncogene c-myc and the cell cycle regulating genes cyclin A and E. Ep-CAM mediates Ca2+-independent homotypic interactions. Formation of Ep-CAM-mediated adhesions have a negative regulatory effect on adhesions mediated by classic cadherins, which may have strong effects on the differentiation and growth of epithelial cells. Ep-CAM overexpression was suggested to be associated with enhanced epithelial proliferation. Ep-CAM is highly expressed in human carcinomas, and is a marker for tumors of epithelial lineage. Ep-CAM is expressed on baso-lateral cell surface in most simple epithelia and many carcinoma types. Also, Ep-CAM reportedly distinguishes adenocarcinomas from pleural mesotheliomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support