Search
Having difficulties with your experiment?
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
Beginning your experiment?
Visit our
Generally, we do not recommend freezing Dynabeads® magnetic beads, as freezing and thawing them may create cracks on the surface of the beads that result in release of iron from the beads and contamination of the sample. Repeated freeze/thaw cycles will enhance this effect and must be avoided. This is all the more important for Dynabeads® magnetic beads with antibodies coated to the surface as freezing of these beads may compromise the performance of the antibodies coated on the bead surface. We recommend storing Dynabeads® magnetic beads at 2 to 8 degrees C in an upright position to ensure that the beads are covered with buffer (drying will reduce their performance). It is always important to resuspend the beads completely and wash them properly before use.
Please review the following possibilities for why your Dynabeads® magnetic beads are not pelleting:
Try these suggestions:
The streptavidin molecule is covalently attached to the surface of the beads; under normal, recommended conditions, negligible leakage is detected (less than 0.2% of total attached streptavidin after 2 months at 37°C). However, it should be noted that not all of the four streptavidin subunits are covalently coupled to the beads. Typically, one or two of the subunits are covalently coupled. Streptavidin is like other proteins; if heated, it can denature and dissociate into subunits. If Dynabeads® Streptavidin magnetic beads are, for instance, boiled, some of the streptavidin subunits may be released (as monomers or aggregates) from the beads. The covalently bound streptavidin subunits will not be affected by such treatment. When streptavidin is bound to biotin, the streptavidin-biotin complex is more stable than the unbound streptavidin molecule.
The binding capacity of streptavidin-coupled Dynabeads® magnetic beads is fragment length-dependent. Reduced binding capacity for large DNA or RNA fragments may be due to steric hindrance. For large DNA or RNA fragments (greater than 2 kb in size), we recommend using Dynabeads® KilobaseBINDER™ Kit. Here are some suggestions for optimizing binding capacity.
Maintain a high pH and a low salt concentration, which will help to maintain the negative charge on both the nucleic acids and the beads.
The background might be caused by nonspecific binding to the BSA on the bead surface. Alternatively, high background might be caused by nonspecific binding to streptavidin. Increasing either the pH or the salt concentration might help reduce the binding. Dynabeads® M-270 Streptavidin magnetic beads might be a better alternative; these beads are not coated with BSA and are hydrophilic, as they are based upon carboxylic acid chemistry.
Dynabeads® M-270 Streptavidin magnetic beads and Dynabeads® MyOne™ Streptavidin C1 magnetic beads have a negatively charged surface. The surface charge of the beads may in some samples cause the beads to float or become sticky or aggregate. The stickiness may be due to electrostatic interactions between the beads or between the beads and the tube wall. Usually we recommend washing the beads in a nonionic detergent like Tween® 20 detergent before doing the experiment. The problem is usually reduced or eliminated by simply adding Tween® 20 detergent to a final concentration of up to 0.1% to the beads, followed by resuspension and washing in buffer without the detergent. An incubation in the Tween® 20 solution may be needed, e.g. 5–10 minutes at room temperature on a roller. In addition, we recommend using siliconized tubes. This treatment will most likely reduce the electrostatic potential of the beads.
We do not recommend this as streptavidin becomes hydrophobic and aggregates during denaturation.
Ribosomal RNA is effectively eliminated by reextracting the mRNA from the eluate. Reuse the same Dynabeads® Oligo(dT)25 beads that were used for the original isolation. Wash the beads twice in Washing Buffer B. Dilute the eluted mRNA with 4 times its volume of Lysis/Binding Buffer, then add the beads. Incubate with mixing at room temperature for 3–5 minutes, then continue with the Direct mRNA Isolation Protocol.
There are several reasons why DNA contamination may occur:
PCR may be inhibited by inhibitors, or an insufficient number of PCR cycles might have been used. For inhibitors, ensure that the washing buffer is brought to room temperature for use, and add the buffer more vigorously. You can also ensure that the supernatant is completely removed at each washing step, or introduce an additional washing step.
We recommend eluting the DNA in Resuspension Buffer, water, or low ionic strength buffer by incubation at 65 degrees C for >5 mins. The complex must be fully resuspended before elution. The elution buffer can be pre-heated to 65 degrees C. After incubation, place tube in magnetic stand for 30 secs and transfer supernatant to a clean tube. To determine the elution efficiency, you can run some of the eluted DNA on an agarose gel. Alternatively, you can perform a PCR with the Dynabeads® to determine if there is residual DNA remaining on the Dynabeads®. Using more pre-heated elution buffer would be helpful to improve the elution efficiency.
High PCR background could be caused by too high a concentration of template DNA, primers, Mg2+, or dNTPs. To fix this, reduce the amount of template DNA, primers, enzyme, and Mg2+ used, elute the DNA prior to PCR amplification, and/or try performing hot-start PCR. High background can also be caused by contamination. Ensure that the supernatant is completely removed at each washing step to avoid carryover.
We suggest checking the white blood cell count, and/or increasing the sample quality and quantity.
You can try to pipette for longer, or move the tube rapidly over an uneven surface, e.g., an iron thread tube rack, to achieve a shaking motion. You can also try to use a tip with a smaller aperture.
Here are some suggestions you can try:
Here are some suggestions you can try:
The biggest cause of yellowing is the addition of isopropanol when Lysis/Binding Buffer is used to make Wash Buffer 1. The use of poor quality isopropanol (not molecular grade) will cause rapid yellowing. High quality alcohols do not cause this.
For Research Use Only. Not for use in diagnostic procedures.