Search
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | |
| Niton Apollo | Niton XL5 Plus | Niton XL3t GOLDD+ | Niton XL2 Plus | Niton XL2 100G | Niton DXL | |
| Analytical Specifications | ||||||
| Technology | LIBS | XRF | XRF | XRF | XRF | XRF |
| Analytical Source | 1064nm laser, charge coupled device (CCD) | 5W x-ray tube, extra large area silicon drift detector (SDD) | 2W x-ray tube, silicon drift detector (SDD) | 2W x-ray tube, silicon drift detector (SDD) | 2W x-ray tube, Si-PIN detector | 2W x-ray tube, Si-PIN detector |
| Elemental Range1 | C, Al, Si, Ti, V, Cr, Mn, Fe, Co, Ni, Cu, Nb, Mo, W | Mg-U (ultra low light element detection), Na (spectrum based detection) | Mg-U | Mg-U | Ti-Bi | Ti-Pb |
| Spot Size | 50 µm standard | 8 mm standard | 8 mm standard | 8 mm standard | 8 mm standard | 8 mm standard |
| Instrument Specifications | ||||||
| Weight2 | 6.4 lbs. | 2.8 lbs. | 3.4 lbs. | 3.4 lbs. | 3.4 lbs. | 17 lbs. |
| Dimensions | 12.0 x 13.0 x 4.0 in. | 9.5 x 8.2 x 2.7 in. | 9.6 x 9.0 x 3.8 in. | 10.0 x 10.8 x 4.0 in. | 10.0 x 10.8 x 4.0 in. | 15.6 x 8.1 x 10.4 in. |
| Memory Readings Capacity3 | 5,000 | < 130,000 | < 10,000 | < 10,000 | < 10,000 | < 10,000 |
| Features | ||||||
| Display Screen | Tilting | Tilting | Tilting | Fixed Angle | Fixed Angle | Fixed Angle |
| Touchscreen/ Directional Keys | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Micro Camera | ✓ | ✓ | ✓ | ✓ |
| ✓ |
| Macro Camera | ✓ | ✓ | ||||
| Bluetooth4 | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| GPS | ✓ | ✓ | ✓ | |||
| WiFi | ✓ | ✓ | ||||
| Hot Swap Battery | ✓ | ✓ | ✓ | |||
Splash + Dust Proof | ✓ | ✓ | ✓ | Coming Soon! | ||
| Detector Protection | N/A | ✓ | ✓ | |||
This table is non-exhaustive and details best sellers from the Niton XRF and LIBS product lines. Instrument specifications and applications vary by model type. Additional calibrations and configurations are available. Speak with a sales representative to learn more.
1. Elemental range dependent upon selected calibration modes. 2. Displayed weight includes battery. 3. Includes spectra. Fewer readings can be saved if macro/ micro images are included. 4. Enables connection to PC, printer, barcode reader, and external GPS.
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | |
| Niton Apollo | Niton XL5 Plus | Niton XL3t GOLDD+ | Niton XL2 Plus | Niton XL2 100G | Niton DXL | |
| Metals / Alloys | ||||||
| Carbon Steel, Low Alloy Steel | ★★★ | ★★ | ★ | ★ | ||
| Stainless Steel L-Grades | ★★★ | |||||
| Scrap Metal Recycling | ★ | ★★ | ★★ | ★★★ | ★★ | |
| Positive Material ID (PMI) | ★★★ | ★★★ | ★★ | ★★ | ★ | |
| Manufacturing QA / QC | ★★ | ★★★ | ★★ | ★★ | ★ | |
| Coatings | ||||||
| Metal Coatings | ★★★ | ★★ | ★ | |||
| Precious Metals | ||||||
| Jewelry & Precious Metals Analysis | ★★ | ★★ | ★★ | ★★★ | ||
| Automotive Catalysts |
| ★★★ |
|
| ||
| Mining | ||||||
| Greenfield Exploration | ★★★ | ★★ | ★ | |||
| Oil & Gas Exploration | ★★★ | ★★ |
| |||
| Grade Control | ★★★ | ★★ | ★ |
| ||
| Rare Earth Elements | ★★★ | |||||
| Environmental Hazards | ||||||
| Soils | ★★★ | ★★ | ★ | |||
| Industrial Lead Paint | ★★ | ★★ | ★ | |||
| Residential Lead Paint | ||||||
| Dust, Air Filters |
| ★★ |
| |||
| Consumer Goods | ||||||
| Children's Toys, Apparel, Jewelry, Furniture | ★★★ | ★★ | ★ | |||
| RoHS / Halogen-free WEEE | ★★★ | ★★ | ★ | |||
| Art & Archaeometry | ||||||
| Metal Artifacts, Pigments, Ceramics, etc.. | ★★★ | ★★ | ★ | |||
★ = Good, ★★ = Better, ★★★ = Best; Rating system evaluates analytical performance, instrument form factor, specifications and application needs.
This table is non-exhaustive and details best sellers from the Niton XRF and LIBS product lines. Instrument specifications and applications vary by model type. Additional calibrations and configurations are available. Speak with a sales representative to learn more.
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | |
| Niton Apollo | Niton XL5 Plus | Niton XL3t GOLDD+ | Niton XL2 Plus | Niton XL2 100G | Niton DXL | |
| Analytical Specifications | ||||||
| Technology | LIBS | XRF | XRF | XRF | XRF | XRF |
| Analytical Source | 1064nm laser, charge coupled device (CCD) | 5W x-ray tube, extra large area silicon drift detector (SDD) | 2W x-ray tube, silicon drift detector (SDD) | 2W x-ray tube, silicon drift detector (SDD) | 2W x-ray tube, Si-PIN detector | 2W x-ray tube, Si-PIN detector |
| Elemental Range1 | C, Al, Si, Ti, V, Cr, Mn, Fe, Co, Ni, Cu, Nb, Mo, W | Mg-U (ultra low light element detection), Na (spectrum based detection) | Mg-U | Mg-U | Ti-Bi | Ti-Pb |
| Spot Size | 50 µm standard | 8 mm standard | 8 mm standard | 8 mm standard | 8 mm standard | 8 mm standard |
| Instrument Specifications | ||||||
| Weight2 | 6.4 lbs. | 2.8 lbs. | 3.4 lbs. | 3.4 lbs. | 3.4 lbs. | 17 lbs. |
| Dimensions | 12.0 x 13.0 x 4.0 in. | 9.5 x 8.2 x 2.7 in. | 9.6 x 9.0 x 3.8 in. | 10.0 x 10.8 x 4.0 in. | 10.0 x 10.8 x 4.0 in. | 15.6 x 8.1 x 10.4 in. |
| Memory Readings Capacity3 | 5,000 | < 130,000 | < 10,000 | < 10,000 | < 10,000 | < 10,000 |
| Features | ||||||
| Display Screen | Tilting | Tilting | Tilting | Fixed Angle | Fixed Angle | Fixed Angle |
| Touchscreen/ Directional Keys | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| Micro Camera | ✓ | ✓ | ✓ | ✓ |
| ✓ |
| Macro Camera | ✓ | ✓ | ||||
| Bluetooth4 | ✓ | ✓ | ✓ | ✓ | ✓ | ✓ |
| GPS | ✓ | ✓ | ✓ | |||
| WiFi | ✓ | ✓ | ||||
| Hot Swap Battery | ✓ | ✓ | ✓ | |||
Splash + Dust Proof | ✓ | ✓ | ✓ | Coming Soon! | ||
| Detector Protection | N/A | ✓ | ✓ | |||
This table is non-exhaustive and details best sellers from the Niton XRF and LIBS product lines. Instrument specifications and applications vary by model type. Additional calibrations and configurations are available. Speak with a sales representative to learn more.
1. Elemental range dependent upon selected calibration modes. 2. Displayed weight includes battery. 3. Includes spectra. Fewer readings can be saved if macro/ micro images are included. 4. Enables connection to PC, printer, barcode reader, and external GPS.
![]() | ![]() | ![]() | ![]() | ![]() | ![]() | |
| Niton Apollo | Niton XL5 Plus | Niton XL3t GOLDD+ | Niton XL2 Plus | Niton XL2 100G | Niton DXL | |
| Metals / Alloys | ||||||
| Carbon Steel, Low Alloy Steel | ★★★ | ★★ | ★ | ★ | ||
| Stainless Steel L-Grades | ★★★ | |||||
| Scrap Metal Recycling | ★ | ★★ | ★★ | ★★★ | ★★ | |
| Positive Material ID (PMI) | ★★★ | ★★★ | ★★ | ★★ | ★ | |
| Manufacturing QA / QC | ★★ | ★★★ | ★★ | ★★ | ★ | |
| Coatings | ||||||
| Metal Coatings | ★★★ | ★★ | ★ | |||
| Precious Metals | ||||||
| Jewelry & Precious Metals Analysis | ★★ | ★★ | ★★ | ★★★ | ||
| Automotive Catalysts |
| ★★★ |
|
| ||
| Mining | ||||||
| Greenfield Exploration | ★★★ | ★★ | ★ | |||
| Oil & Gas Exploration | ★★★ | ★★ |
| |||
| Grade Control | ★★★ | ★★ | ★ |
| ||
| Rare Earth Elements | ★★★ | |||||
| Environmental Hazards | ||||||
| Soils | ★★★ | ★★ | ★ | |||
| Industrial Lead Paint | ★★ | ★★ | ★ | |||
| Residential Lead Paint | ||||||
| Dust, Air Filters |
| ★★ |
| |||
| Consumer Goods | ||||||
| Children's Toys, Apparel, Jewelry, Furniture | ★★★ | ★★ | ★ | |||
| RoHS / Halogen-free WEEE | ★★★ | ★★ | ★ | |||
| Art & Archaeometry | ||||||
| Metal Artifacts, Pigments, Ceramics, etc.. | ★★★ | ★★ | ★ | |||
★ = Good, ★★ = Better, ★★★ = Best; Rating system evaluates analytical performance, instrument form factor, specifications and application needs.
This table is non-exhaustive and details best sellers from the Niton XRF and LIBS product lines. Instrument specifications and applications vary by model type. Additional calibrations and configurations are available. Speak with a sales representative to learn more.
Interested in learning how XRF technology works? Download the eBook to discover the science behind x-ray fluorescence and how it can perform for your business. Developed by our team of experts, you’ll learn the basics, industries that may benefit from handheld XRF and much more!
Want to learn how LIBS technology works? Download the eBook to discover the science behind laser induced breakdown spectroscopy. Developed by our team of experts, you’ll learn what businesses can benefit from portable LIBS, how the technology works, and what to look for when purchasing a LIBS analyzer- plus much more!
Metals|Mining
Metals|Mining
Metals|Mining
Metals|Mining