Search
Nalgene volumetric flasks are a safer alternative to glass. They won’t break, chip or shatter amid the bustle and bumps of everyday lab work. Enjoy their light weight and ease of handling. Simplify volume readings: liquid in plastic volumetrics don’t form a meniscus, so level liquid avoids confusion and errors. Each flask is individually hand calibrated, engraved and inked with a volume fill line to provide precision and ASTM E228/ISO 384 Class B accuracy for the life of the product.
Trust Thermo Scientific Nalgene volumetric flasks to be made from the highest quality laboratory-grade polypropylene copolymer (PPCO) and polymethylpentene (PMP) materials for dependably low extractables across a wide variety of lab applications. Choose polypropylene for its low cost and good chemical compatibility with a wide range of lab chemicals. Get improved chemical resistance and clarity, too, with PMP. Enjoy leak-free performance with our Nalgene leakproof* guarantee.
| Nalgene PPCO Volumetric Flask with Screw Cap | Nalgene PMP Volumetric Flask with Screw Cap |
| Lower cost, milky white, good chemical resistance | Better clarity and chemical resistance |
![]() | ![]() |
| Screw-on cap | Screw-on cap |
| Engraved and inked fill line | Engraved and inked fill line |
| 50mL–1000mL | 50mL–1000mL |
| PPCO flask w/PP closure | PMP flask w/PP closure |
| Milky white translucent | Light white haze |
| Meets accuracy requirements of ASTM E288 and ISO 384, Class B | Meets accuracy requirements of ASTM E288 and ISO 384, Class B |
| Calibrated TC/TD at 20oC | Calibrated TC/TD at 20oC |
| Autoclavable, but will affect accuracy | Autoclavable, but will affect accuracy |
| Complies with Part 21 CFR 177.1520 for food use | No food compliance |
| BPA-Free | BPA-Free |
| Phthalate-free | Phthalate-free |
| Nalgene PPCO Volumetric Flask with Screw Cap | Nalgene PMP Volumetric Flask with Screw Cap | |
| Acids: dilute or weak | E | E |
| Acids^: strong, concentrated | G | E |
| Alcohols, aliphatic | E | E |
| Aldehydes | G | G |
| Bases/alkali | E | E |
| Esters | G | E |
| Hydrocarbons, aliphatic | G | G |
| Hydrocarbons, aromatic | N | N |
| Hydrocarbons, halogenated | N | N |
| Ketones, aromatic | N | F |
| Oxidizing agents, strong | F | G |
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
| E | No damage after 30 days constant exposure at 20 oC. |
| G | Little or no damage after 30 days constant exposure at 20oC. |
| F | Some effect after 7 days constant exposure at 20oC. |
| N | Not recommended. Immediate damage may occur. |
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Nalgene bottles and closures under conditions of their planned applications. Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^Except for oxidizing acids, see”Oxidizing agents, strong”
For a more detailed chemical resistance guide by chemical and application temperature, please refer to the Nalgene Chemical Resistance Wall Chart or contact
technicalsupport@thermofisher.com.