Search
At Thermo Fisher Scientific, we understand that harvest and clarification have become more demanding as bioprocessing shifts toward higher cell densities, higher titers, and evolving feed strategies. These conditions can increase the amount and variability of solids that must be addressed during clarification. Clarification strategies are evaluated early to anticipate differences in solids load, viscosity, and impurity profiles that emerge during upstream processing. Depth filtration solutions are one of several clarification approaches used to remove cells, cell debris, and fine insoluble particulates.
Depth filtration solutions, including Zeta Plus depth filters, are available in capsule, cartridge, and sheet formats. These solutions enable clarification strategies to be applied across process development and manufacturing using the same underlying media. Format selection typically reflects process scale, facility design, handling preferences, and system closure requirements. As processes progress, formats may be reevaluated to align with manufacturing needs.
Depth filtration media and layer configurations should align with process-specific solids load, impurity profiles, and throughput requirements. Media characteristics and single- or dual-layer designs are considered when evaluating contaminant holding capacity and impurity retention within the filter structure.
Depth filtration portfolios also include application-specific media designed to address clarification requirements beyond standard harvest workflows.
| Media | Course | Fine |
| ZB | N/A | 60ZB05A, 90ZB08A, 120ZB08A |
| SP | 05SP01A, 10SP02A, 30SP02A | 60SP02A, 60SP03A, 60SP05A, 90SP05 |
| LA | 30LA | 60LA, 90LA |
In many bioprocesses, depth filtration is introduced during harvest and clarification as either a primary clarification step or a secondary clarification step. It is commonly evaluated following centrifugation or similar separation steps to manage remaining cells and insoluble material. Placement and configuration are determined during process development, taking into account harvest composition, facility constraints, and how clarification interfaces with downstream operations.
Format selection usually depends on how a process is operated at a given stage rather than on a single configuration. During development or scale-up, single-use capsules may be evaluated for flexibility, while cartridges or sheets are often used in facilities with existing reusable infrastructure. Scale, handling approach, and integration with surrounding unit operations are reviewed during process design.
High-density cultures can introduce higher solids levels, increased debris, and changes in fluid viscosity that affect clarification behavior. Impurity profiles may also shift as titers increase, influencing how media interact with process streams. During process development, these factors are evaluated together to inform media grade and layer configuration. In some cases, these factors may prompt exploration of modified clarification strategies to accommodate high-titer process demands.
Depth filtration can support scale-up when clarification strategies are explored early and revisited as processes advance. Evaluating the same media across scalable formats allows development-stage learnings to inform later configuration decisions. Format selection and media choices are commonly reassessed during tech transfer to align clarification steps with manufacturing-scale equipment and downstream process requirements.
Depth filtration may be combined with additional clarification or polishing technologies when process streams present higher impurity loads or increased variability. In these cases, depth filtration can be paired with approaches, such as chromatographic clarification or membrane-based steps, to further condition the stream. Decisions to combine technologies are typically made during process development based on impurity profiles, downstream sensitivity, and overall workflow design.
Intended use of the products mentioned on this page vary. For specific intended use statements, please refer to the Instructions for Use for the product.