Search
shRNA Vectors for RNAi |
Short hairpin RNAs (shRNAs) are a vector-based solution used to silence gene expression via RNA interference (RNAi) pathways. Our shRNA vectors are designed to deliver robust and stable gene silencing through the expression of shRNAs, which are subsequently processed into small interfering RNAs (siRNAs) within the cell, leading to targeted mRNA degradation and eventually suppressing protein expression. We have a diverse range of shRNA vectors, each optimized for efficient and reliable knockdown of specific genes. Whether your experiment requires constitutive or inducible expression, our vectors are equipped with various promoters and selection markers to suit experimental needs.
A vector expressing the shRNA is generated by designing a short, double-stranded DNA oligo encoding a sense-loop-antisense sequence to the targeted gene. On either side, this shRNA sequence will have a four-nucleotide overhang that is compatible with the entry vector construct and can be cloned into the pENTR/H1/TO or pENTR/U6 entry vector via a brief benchtop ligation reaction.
The entry vector contains the U6 Pol III-type promoter and the Pol III terminator sequence. Once the double-stranded oligo sequence is cloned into the vector, an RNAi cassette that expresses the shRNA sequence will be created.
A vector expressing the shRNA is generated by designing a short double-stranded DNA oligo containing a sense-loop-antisense sequence against the target. The shRNA sequence has 4-nucleotide overhangs on the ends that can be ligated into the pENTR/H1/TO or pENTR/U6 entry vector via a brief benchtop reaction. The entry vector contains either the H1 promoter with two tetracycline operator sites flanking the TATA region, or the U6 Pol III type promoter and the Pol III terminator sequence. Cloning the double-stranded oligo into the vector creates an RNAi cassette that expresses the shRNA sequence.
Transfer your shRNA sequence into one or more of the BLOCK-iT destination (DEST) vectors to get efficient delivery and expression of shRNA in mammalian cells. Each DEST vector is designed with Gateway cloning to easily transfer the RNAi cassette from the BLOCK-iT pENTR vector to a BLOCK-iT-DEST vector in just one hour. Choose what vector to use depending on cell type and selection needs. Get delivery in virtually any mammalian cell type, including hard-to-transfect, primary, and even non-dividing cell types using:
Choose a shRNA vector based on applications and advantages.
| Vector type | Attributes | Application | Advantages | Products |
|---|---|---|---|---|
| BLOCK-iT Entry Vectors | Vector: pENTR/U6 System: plasmid Selection: none Promoter: U6 | Transient RNAi in transfectable mammalian cells |
| U6 RNAi Entry Vector Kit |
| Viral BLOCK-iT DEST Vectors | Vector: pLenti4/BLOCK-iT-DEST System: lentiviral Selection: zeocin Promoter: none | Stable RNAi in virtually any mammalian cell type or animal model |
| Lentiviral RNAi Zeo Gateway Vector Kit |
| Vector: pLenti6/BLOCK-iT-DEST System: lentiviral Selection: blasticidin Promoter: none | Lentiviral |
| Lentiviral RNAi Gateway Vector Kit Lentiviral RNAi Expression System | |
| Vector: pAd/BLOCK-iT-DEST System: adenoviral Selection: none Promoter: none | Adenoviral |
| Adenoviral RNAi Expression System | |
| Support vectors | Vector: pLenti6/TR System: lentiviral Selection: blasticidin Promoter: CMV | To generate stable cell lines that constitutively express the tetracycline repressor (TR) |
| pLenti6/TR Vector Kit |
| Vector: pcDNA6/TR System: plasmid Selection: blasticidin Promoter: CMV | Plasmid |
| pcDNA 6/TR |
Technical inquires:
Our Technical Application Scientists are available to help assist you at techsupport@thermofisher.com
Ordering & Order Status inquires:
If you have questions about pre-designed RNAi orders and order status, please contact us at genomicorders@thermofisher.com
If you have any questions about Custom RNAi orders and order status, please contact us at RNAiSupport@thermofisher.com
For Research Use Only. Not for use in diagnostic procedures.